Skip to Content
Merck
All Photos(1)

Key Documents

44944

Sigma-Aldrich

Iron(III) chloride hexahydrate

puriss. p.a., ACS reagent, crystallized, 98.0-102% (RT)

Synonym(s):

Ferric chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeCl3 · 6H2O
CAS Number:
Molecular Weight:
270.30
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Assay:
98.0-102% (RT)
form:
crystals or chunks

grade

ACS reagent
puriss. p.a.

Quality Level

vapor pressure

1 mmHg ( 194 °C)

Assay

98.0-102% (RT)

form

crystals or chunks

quality

crystallized

impurities

≤0.001% total nitrogen (N)
≤0.002% free chlorine (Cl)
≤0.01% phosphorous compounds (as PO4)

bp

280-285 °C (lit.)

mp

37 °C (lit.)

anion traces

nitrate (NO3-): ≤100 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤10 mg/kg
Co: ≤50 mg/kg
Cr: ≤100 mg/kg
Cu: ≤20 mg/kg
Fe2+: ≤20 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤1000 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤20 mg/kg
Zn: ≤20 mg/kg

SMILES string

O.O.O.O.O.O.Cl[Fe](Cl)Cl

InChI

1S/3ClH.Fe.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

NQXWGWZJXJUMQB-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Iron(III) chloride hexahydrate can be obtained from crystallization of iron(III) chloride from water. Three-dimensional single-crystal X-ray-diffraction studies of its crystal structure suggest its crystals are monoclinic and C2h3-2/m space group.

Application

Iron(III) chloride hexahydrate (FeCl3.6H2O) may be employed as an oxidant for the following conversions:
  • Synthesis of 2-substituted 4(3H)-quinazolinones (yield = 77-93%).
  • Oxidation of 1,4-dihydropyridines (1,4-DHPs) to corresponding pyridines.
  • Oxidation of 4-isopropyl-1,4-DHP to dealkylated pyridine.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benign and efficient synthesis of 2-substituted 4(3H)-quinazolinones mediated by iron (III) chloride hexahydrate in refluxing water.
Wang G-W, et al.
Bulletin of the Chemical Society of Japan, 79(9), 1426-1430 (2006)
Amlan Chakraborty et al.
International journal of molecular sciences, 21(5) (2020-03-04)
Despite developments in pulmonary radiotherapy, radiation-induced lung toxicity remains a problem. More sensitive lung imaging able to increase the accuracy of diagnosis and radiotherapy may help reduce this problem. Super-paramagnetic iron oxide nanoparticles are used in imaging, but without further
Crystal structure of ferric chloride hexahydrate.
Lind MD.
J. Chem. Phys. , 47(3), 990-993 (1967)
Ferric chloride hexahydrate: a convenient reagent for the oxidation of hantzsch 1, 4-dihydropyridines.
Lu J, et al.
Synthetic Communications, 31(17), 2625-2630 (2001)
Rajendra Upadhya et al.
mBio, 9(6) (2018-11-22)
Chitin is an essential component of the cell wall of Cryptococcus neoformans conferring structural rigidity and integrity under diverse environmental conditions. Chitin deacetylase genes encode the enyzmes (chitin deacetylases [Cdas]) that deacetylate chitin, converting it to chitosan. The functional role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service