Skip to Content
Merck
All Photos(2)

Key Documents

C4386

Sigma-Aldrich

Activated charcoal

greener alternative

acid-washed with hydrochloric acid

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52
Pricing and availability is not currently available.

vapor pressure

<0.1 mmHg ( 20 °C)

Quality Level

form

powder

autoignition temp.

842 °F

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

impurities

≤10% moisture content

mp

3550 °C (lit.)

anion traces

chloride (Cl-): ≤0.1%

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Activated charcoal, acid-washed with hydrochloric acid forms a composite with natural cellulose, which can be used in the fabrication of a flexible electrode with a specific capacitance of ~103 Fg-1 for supercapacitor application.[1] It can also be used in the preparation of ultracapacitors with asymmetric configuration.[2]
Used in a wide variety of biochemical and immunological applications to remove reactants and waste products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Power loss and energy density of the asymmetric ultracapacitor loaded with molybdenum doped manganese oxide
Wang Y, et al.
Electrochimica Acta, 68, 95-102 (2012)
Cellulose and activated carbon based flexible electrical double-layer capacitor electrode: Preparation and characterization
Murashko K, et al.
Energy, 119, 435-441 (2017)
Milk Progesterone ELISA to Monitor the Success of Artificial Insemination (AI) in Buffaloes.
Kabir MS, et al.
International Journal of BioResearch, 1(3), 5-9 (2011)
Kohsuke Ohmatsu et al.
Nature chemistry, 6(1), 47-51 (2013-12-19)
The development of a general catalytic method for the direct and stereoselective construction of contiguous all-carbon quaternary stereocentres remains a formidable challenge in chemical synthesis. Here, we report a highly enantio- and diastereoselective [3+2] annulation reaction of 5-vinyloxazolidinones and activated
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service