Skip to Content
Merck
All Photos(1)

Key Documents

71380

Sigma-Aldrich

Sodium chloride

puriss. p.a., ≥99.5% (AT)

Synonym(s):

Halite, NaCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals
solubility:
water: soluble
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 865 °C)

Quality Level

grade

puriss. p.a.

Assay

≥99.5% (AT)

form

powder or crystals

impurities

≤0.001% total nitrogen (N)

pH

5.0-8.0 (20 °C, 50 mg/mL in H2O)

mp

801 °C (lit.)

solubility

water: soluble

anion traces

bromide (Br-): ≤50 mg/kg
iodide (I-): ≤10 mg/kg
phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤1 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

[Na+].[Cl-]

InChI

1S/ClH.Na/h1H;/q;+1/p-1

InChI key

FAPWRFPIFSIZLT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium chloride (NaCl) is a colorless solid and its crystals belong to the cubic crystal system. It is widely employed preservative for meat, vegetable and fish products. NaCl is a useful precursor for the production of sodium and chlorine containing compounds.[1] Its effect on the growth of salmonellae (mixed inoculum of Salmonella thompson, S. stanley and S. infantis) has been investigated.[2]

Application

Sodium chloride was used in the isolation medium along with other constituents, prepared for incubating bone-marrow cells, during the isolation and expansion of hBM-MSCs (human bone-marrow mesenchymal stem cells).[3]

Other Notes

Sales restrictions may apply

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana B Faia-Torres et al.
Biomaterials, 35(33), 9023-9032 (2014-08-12)
Tissue engineering using scaffold-cell constructs holds the potential to develop functional strategies to regenerate bone. The interface of orthopedic implants with the host tissues is of great importance for its later performance. Thus, the optimization of the implant surface in
Tianji Ma et al.
ACS applied materials & interfaces, 11(13), 12578-12585 (2019-03-13)
The salinity gradient between brine and fresh water is an abundant source of power which can be harvested by two major membrane methods: pressure-retarded osmosis and reversed electrodialysis. Nowadays, the latter technology is close to real application, but it still
Tianji Ma et al.
Nanomaterials (Basel, Switzerland), 11(1) (2021-01-23)
The modification of the inner nanopore wall by polymers is currently used to change the specific properties of the nanosystem. Among them, the polyethylene glycol (PEG) is the most used to prevent the fouling and ensure the wettability. However, its
A M Gibson et al.
International journal of food microbiology, 6(2), 155-178 (1988-03-01)
The growth responses of salmonellae (mixed inoculum of Salmonella thompson, S. stanley and S. infantis) as affected by NaCl concentration, pH level and storage temperature were studied in laboratory medium. Growth curves were obtained at 5 concentrations of NaCl (0.5-4.5%
Eagleson M.
Concise Encyclopedia Chemistry, 996-996 (1994)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service