Skip to Content
Merck
All Photos(1)

Key Documents

673838

Sigma-Aldrich

Petroleum ether

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
grade:
ACS reagent
bp:
30-60 °C (lit.)
35-60 °C
vapor pressure:
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

grade

ACS reagent

Quality Level

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

impurities

acidity, passes test

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)
35-60 °C

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether is a fraction of petroleum that contains aliphatic hydrocarbons. It serves as a laboratory solvent because of its low boiling point and non-polar characteristics.

Application

Petroleum ether can be used as a solvent for extracting:
  • Soluble compounds from the mushroom samples.
  • Carotenoids from the marine Thraustochytrium sp.ONC-T18.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F

Flash Point(C)

-49 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S C Bondy et al.
Archives of toxicology, 69(6), 410-414 (1995-01-01)
Levels of glutathione and activity of glutamine synthetase were assayed in organs of rats following inhalation of a heterogeneous solvent mixture containing both aliphatic and aromatic hydrocarbons. This mixture was administered for 3 weeks (6 h daily) at two levels
I P Dick et al.
Human & experimental toxicology, 16(11), 645-651 (1998-01-14)
1. The absorption and distribution of lindane through skin was examined using human volunteers. Two different preparations were employed, one with acetone as the vehicle and the other, a commercial product, consisting primarily of white spirit as the solvent base.
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
A Steensgaard et al.
Neurotoxicology, 17(3-4), 785-792 (1996-10-01)
Several epidemiological studies of workers occupationally exposed to white spirit show that neuropsychiatric disorders are a frequent cause of early disability pension in this population compared with non-exposed controls. In the rat, we have demonstrated that exposure to different kinds
I P Dick et al.
Human & experimental toxicology, 16(11), 652-657 (1998-01-14)
1. The absorption of lindane through human skin was assessed in vitro using static diffusion cells with 50% aqueous ethanol as the receptor fluid. Four formulations of lindane were applied, three of which were commercial preparations. The dermal distribution of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service