Skip to Content
Merck
All Photos(1)

Key Documents

320447

Sigma-Aldrich

Petroleum ether

ACS reagent

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12191502
NACRES:
NA.07
grade:
ACS reagent
bp:
30-60 °C (lit.)
35-60 °C
vapor pressure:
25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

grade

ACS reagent

vapor density

2.5 (vs air)

vapor pressure

25.8 psi ( 55 °C)
7.99 psi ( 20 °C)

form

liquid

autoignition temp.

475 °F

expl. lim.

8 %

dilution

(for analytical testing)

impurities

acidity, passes test

evapn. residue

≤0.001%

color

APHA: ≤10

refractive index

n20/D 1.363 (lit.)

bp

30-60 °C (lit.)
35-60 °C

density

0.64 g/mL at 25 °C (lit.)

SMILES string

[Mg+]c1ccc(cc1)C.[Br-]

InChI

1S/C7H7.BrH.Mg/c1-7-5-3-2-4-6-7;;/h3-6H,1H3;1H;/q;;+1/p-1

InChI key

ZRJNGFJIBZKXTP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Petroleum ether is the petroleum fraction, consisting of C5 and C6 hydrocarbons (Pentanes and hexanes are the major constituents), commonly used as a laboratory solvent due to its low boiling point and non-polar characteristics.

Application

Petroleum ether can be used as a solvent for extracting:
  • Non-polar compounds from various samples, such as plant materials or organic mixtures.
  • Carotenoids from the marine Thraustochytrium sp. ONC-T18.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-56.2 °F

Flash Point(C)

-49 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal concentration and antioxidant activity of edible mushrooms from Turkey
Sarikurkcu C, et al.
Food Chemistry, 175, 549-555 (2015)
Critical assessment of various techniques for the extraction of carotenoids and co-enzyme Q10 from the thraustochytrid strain ONC-T18
Armenta RE, et al.
Journal of Agricultural and Food Chemistry, 54(26), 9752-9758 (2006)
Catherine F Poh et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 12(22), 6716-6722 (2006-11-24)
Genetically altered cells could become widespread across the epithelium of patients with oral cancer, often in clinically and histologically normal tissue, and contribute to recurrent disease. Molecular approaches have begun to yield information on cancer/risk fields; tissue optics could further
S C Bondy et al.
Archives of toxicology, 69(6), 410-414 (1995-01-01)
Levels of glutathione and activity of glutamine synthetase were assayed in organs of rats following inhalation of a heterogeneous solvent mixture containing both aliphatic and aromatic hydrocarbons. This mixture was administered for 3 weeks (6 h daily) at two levels
A Löf et al.
Pharmacology & toxicology, 85(2), 92-97 (1999-09-17)
Petroleum products with low content of aromatics have been increasingly used during the past years. This study investigates tissue disposition of dearomatised white spirit. In addition, brain neurotransmitter concentrations were measured. Male rats were exposed by inhalation to 0, 400

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service