Skip to Content
Merck
All Photos(1)

Key Documents

61230

Sigma-Aldrich

Copper(II) sulfate

puriss. p.a., anhydrous, ≥99.0% (RT)

Synonym(s):

Cupric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4
CAS Number:
Molecular Weight:
159.61
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

vapor pressure

7.3 mmHg ( 25 °C)

grade

puriss. p.a.

Assay

≥99.0% (RT)

form

powder

impurities

≤0.005% total nitrogen (N)

loss

≤1.0% loss on drying, 250 °C

pH

3.5-4.5 (20 °C, 50 g/L)

mp

200 °C (dec.) (lit.)

density

3.603 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Mg: ≤20 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

application(s)

environmental

SMILES string

[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

ARUVKPQLZAKDPS-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II) sulfate (CuSO4) is an anhydrous copper(II) salt. It can be synthesized from CuSO4.5H2O, via dehydration. Copper(II) sulfate adsorbed on silica gel participates in the conversion of secondary and tertiary alcohols to olefins. Copper(II) sulfate-alumina has been employed as a solid-reagent system during the synthesis of benzils.

Application

Copper(II) sulfate (CuSO4) may be employed as metal source in the synthesis of copper oxide (CuO) hollow nanospheres.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annalisa La Gatta et al.
Clinical, cosmetic and investigational dermatology, 13, 87-97 (2020-02-26)
Hyaluronic Acid (HA) fillers are among the most used products in cosmetic medicine. Companies offer different formulations to allow full facial treatment and/or remodeling. Gels are being studied to establish the biophysical properties behind the specific clinical use and a
Copper (II) Sulfate.
Hoffman RV, et al.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2007)
Fabrication of Copper (II) Oxide Hollow Nanosphere Using ABC Block Copolymer Templates and Its Application as Anode Materials in Lithium Ion Batteries.
Alam MM, et al.
Chemistry Letters (Jpn), 43(9), 1426-1428 (2014)
Solid State Oxidation of Benzoins on Alumina-Supported Copper (II) Sulfate under Microwave Irradiation?.
Varma RS.
J. Chem. Res. Synop., 6, 324-325 (1998)
Dehydration of alcohols catalyzed by copper (II) sulfate adsorbed on silica gel.
Nishiguchi T, et al.
Tetrahedron Letters, 28(39), 4565-4568 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service