Skip to Content
Merck
All Photos(3)

Key Documents

C8027

Sigma-Aldrich

Copper(II) sulfate pentahydrate

BioReagent, suitable for cell culture, ≥98%

Synonym(s):

Cupric sulfate pentahydrate, Sulfuric acid, copper(II) salt (1:1) pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CuSO4 · 5H2O
CAS Number:
Molecular Weight:
249.69
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.71

vapor pressure

7.3 mmHg ( 25 °C)

product line

BioReagent

Assay

≥98%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

110 °C (dec.) (lit.)

SMILES string

O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

InChI

1S/Cu.H2O4S.5H2O/c;1-5(2,3)4;;;;;/h;(H2,1,2,3,4);5*1H2/q+2;;;;;;/p-2

InChI key

JZCCFEFSEZPSOG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Copper(II) sulfate pentahydrate is known as blue vitriol. Structurally, in the pentahydrate molecule, each copper(II) ions is surrounded by four water molecules in the corners and the fifth water molecule is attached by hydrogen bonding. Copper (II) sulfate has many applications including preparation of Bordeaux mixture, a fungicide preparation. Electroplating, timber preservation and textile industry use copper (II) sulfate.

Application

Copper(II) sulfate pentahydrate has been used:
  • as an additive in trace element solution preparation in solid glucose minimal media
  • as a component of adamsII solution in Pneumococcal media
  • in the preparation of alginate gel for drug encapsulation

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Dictionary of Chemistry (2016)
Intensification of biopolymeric microparticles production by ultrasonic assisted atomization
Barba AA, et al.
Chemical Engineering and Processing, 48(10), 1477-1483 (2009)
Preparation of Purified Gram-positive Bacterial Cell Wall and Detection in Placenta and Fetal Tissues
Mann B, et al.
Bio-protocol, 6(23) (2016)
Establishing a Biofilm Co-culture of Pseudomonas and Aspergillus for Metabolite Extraction
Zheng H, et al.
Current Biology, 5(23), e1667-e1667 (2015)
Keli Chai et al.
Viruses, 13(1) (2020-12-31)
Membrane proteins of enveloped viruses have been reported to undergo palmitoylation, a post-translational modification often having a critical role in the function of these viral proteins and hence viral replication. In this study, we report that the foamy virus (FV)

Articles

Importance and uses of copper in serum-free eukaryotic, including hybridoma and Chinese Hamster Ovary (CHO), cell cultures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service