Skip to Content
Merck
All Photos(2)

Key Documents

484016

Sigma-Aldrich

Potassium hydroxide

reagent grade, 90%, flakes

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
flakes
solubility:
water: soluble 1120 g/L

grade

reagent grade

Agency

suitable for EPA 1613
suitable for SM 5210

vapor pressure

1 mmHg ( 719 °C)

Assay

90%

form

flakes

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

solubility

water: soluble 1120 g/L

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide (KOH) is a hygroscopic, water soluble base synthesized by the electrolysis of potassium chloride solution. It is the raw material for the synthesis of many potassium compounds. KOH reacts with CO2 to form potassium carbonate. This property can be utilized to absorb CO2. It participates as catalyst in aldol condensation reactions. KOH anisotropic etching of silicon is used for the fabrication of sub-micron silicon nitride waveguides using conventional contact lithography. Pulverized KOH may be used for the preparation of chalcones.

Application

Potassium hydroxide (KOH) was used in the following processes:
  • Quantification of glutathione (GSH) and glutathione disulfide (GSSG) in biological samples by HPLC.
  • Preparation of buffer for use in the Investigation of seed fatty acid (FA) composition, protein, oil and sugar content.
  • Stable isotopic profiling study of Caenorhabditis elegans.
Potassium hydroxide may be employed as an etchant for the laser modified glass. In combination with tellurium powder, it mediates the quantitative pinacolization of aromatic carbonyl compounds.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seed Storage Reserve Analysis
van Erp H, et al.
Bio-protocol, 4(20), e1263-e1263 (2014)
Marni J Falk et al.
Journal of visualized experiments : JoVE, (48)(48), doi:10-doi:10 (2011-03-16)
Stable isotopic profiling has long permitted sensitive investigations of the metabolic consequences of genetic mutations and/or pharmacologic therapies in cellular and mammalian models. Here, we describe detailed methods to perform stable isotopic profiling of intermediary metabolism and metabolic flux in
Eagleson M.
Concise Encyclopedia Chemistry, 888-888 (1994)
Yuewang Huang et al.
Optics express, 23(5), 6780-6786 (2015-04-04)
We demonstrate a novel technique to fabricate sub-micron silicon nitride waveguides using conventional contact lithography with MEMS-grade photomasks. Potassium hydroxide anisotropic etching of silicon facilitates line reduction and roughness smoothing and is key to the technique. The fabricated waveguides is
Khan, R.H. et al.
Synthetic Communications, 27, 2193-2193 (1997)

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service