Skip to Content
Merck
All Photos(3)

Key Documents

306568

Sigma-Aldrich

Potassium hydroxide

semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)

Synonym(s):

Caustic potash

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOH
CAS Number:
Molecular Weight:
56.11
EC Number:
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38100303
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.99% trace metals basis (Purity excludes sodium content.)
grade:
semiconductor grade
form:
pellets
Pricing and availability is not currently available.

grade

semiconductor grade

Quality Level

vapor pressure

1 mmHg ( 719 °C)

Assay

99.99% trace metals basis (Purity excludes sodium content.)

form

pellets

composition

KOH, >85.0%

impurities

15% water

pH

~13.5 (25 °C, 5.6 g/L)

mp

361 °C (lit.)

cation traces

Na: ≤500.0 ppm

SMILES string

[OH-].[K+]

InChI

1S/K.H2O/h;1H2/q+1;/p-1

InChI key

KWYUFKZDYYNOTN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium hydroxide (KOH), also known as caustic potash. It is used as a significant precursor to many potassium containing products.

Application

In combination with tellurium powder, mediates the quantitative pinacolization of aromatic carbonyl compounds.[1]
Potassium hydroxide can be used:
  • As a base in the synthesis of imines, starting from amines and alcohols.
  • As a base in β-alkylation of secondary alcohols with primary alcohols without any transition metal catalyst.
  • In combination with aluminium for pinacolization of aromatic aldehydes and reduction of hindered ketones.
  • As a precipitating agent in the synthesis of MnZn ferrites nanoparticles (Mn0.8Zn0.2Fe2O4 ferrite nanoparticles) by modified hydrothermal method.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qilun Wang et al.
Nature communications, 11(1), 4246-4246 (2020-08-28)
Water electrolysis offers a promising energy conversion and storage technology for mitigating the global energy and environmental crisis, but there still lack highly efficient and pH-universal electrocatalysts to boost the sluggish kinetics for both cathodic hydrogen evolution reaction (HER) and
Ian T McCrum et al.
The journal of physical chemistry. C, Nanomaterials and interfaces, 122(29), 16756-16764 (2018-09-28)
The effect of the alkali-metal cation (Li+, Na+, K+, and Cs+) on the non-Nernstian pH shift of the Pt(554) and Pt(533) step-associated voltammetric peak is elucidated over a wide pH window (1-13), through computation and experiment. In conjunction with our
Haochen Zhang et al.
Nature communications, 10(1), 3340-3340 (2019-07-28)
Electroreduction of carbon dioxide to hydrocarbons and oxygenates on copper involves reduction to a carbon monoxide adsorbate followed by further transformation to hydrocarbons and oxygenates. Simultaneous improvement of these processes over a single reactive site is challenging due to the
Takahiro Naito et al.
ChemSusChem, 13(22), 5921-5933 (2020-09-03)
Hydrogen production from renewable energy and ubiquitous water has a potential to achieve sustainability, although current water electrolyzers cannot compete economically with the fossil fuel-based technology. Here, we evaluate water electrolysis at pH 7 that is milder than acidic and alkaline
Maria D Cabezas et al.
Methods in cell biology, 119, 261-276 (2014-01-21)
The extracellular matrix (ECM) is a complex, spatially inhomogeneous environment that is host to myriad cell-receptor interactions that promote changes in cell behavior. These biological systems can be probed and simulated with engineered surfaces, but doing so demands careful control

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service