Skip to Content
Merck
All Photos(1)

Documents

439185

Sigma-Aldrich

Hexane, mixture of isomers

≥98.5%, suitable for HPLC

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.06

product name

Hexane, mixture of isomers, suitable for HPLC, ≥98.5%

vapor density

3 (vs air)

Quality Level

Assay

≥98.5%

form

liquid

autoignition temp.

437 °F

expl. lim.

7.5 %

technique(s)

HPLC: suitable

impurities

<0.020% water

evapn. residue

<0.0005%

color

APHA: ≤10

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

solubility

ethanol: soluble(lit.)

density

0.672 g/mL at 25 °C (lit.)
0.672 g/mL at 25 °C

λ

H2O reference

UV absorption

λ: 195 nm Amax: 1.00
λ: 210 nm Amax: 0.30
λ: 220 nm Amax: 0.10
λ: 230 nm Amax: 0.05
λ: 240 nm Amax: 0.02
λ: 250-400 nm Amax: 0.01

application(s)

food and beverages

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Meets ACS specifications

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F

Flash Point(C)

-23 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solving the Clogging Problem: Precipitate-Forming Reactions in Flow.
Poe SL, et al.
Angewandte Chemie (International Edition in English), 45(10), 1544-1548 (2006)
Separation of hexane isomers in a metal-organic framework with triangular channels.
Herm ZR, et al.
Science, 340(6135), 960-964 (2013)
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service