Skip to Content
Merck
All Photos(3)

Key Documents

227064

Sigma-Aldrich

Hexane, mixture of isomers

≥99%, anhydrous

Synonym(s):

Hexanes, Isohexane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14
CAS Number:
Molecular Weight:
86.18
Beilstein:
1730733
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
bp:
68-70 °C (lit.)

Product Name

Hexane, mixture of isomers, anhydrous, ≥99%

grade

anhydrous

Quality Level

vapor density

3 (vs air)

Assay

≥99%

form

liquid

autoignition temp.

437 °F

expl. lim.

7.5 %

impurities

<0.001% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.379 (lit.)

bp

68-70 °C (lit.)

mp

−95 °C (lit.)

density

0.672 g/mL at 25 °C (lit.)

SMILES string

C(CCC)(C)C

InChI

1S/C6H14/c1-4-5-6(2)3/h6H,4-5H2,1-3H3

InChI key

AFABGHUZZDYHJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexane, mixture of isomers is a non-renewable organic solvent mainly used for the dissolution and extraction of oils and fats.

Application

Hexane, mixture of isomers may be used as a carrier phase for microfluidic devices.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-9.4 °F - closed cup

Flash Point(C)

-23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Alternative bio-based solvents for extraction of fat and oils: solubility prediction, global yield, extraction kinetics, chemical composition and cost of manufacturing.
Sicaire A-G, et al.
International Journal of Molecular Sciences, 16(4), 8430-8453 (2015)
Solving the Clogging Problem: Precipitate-Forming Reactions in Flow.
Poe SL, et al.
Angewandte Chemie (International Edition in English), 45(10), 1544-1548 (2006)
Emily N Abada et al.
Biomedical microdevices, 20(4), 86-86 (2018-10-07)
While extracorporeal membrane oxygenation (ECMO) is a valuable therapy for patients with lung or heart failure, clinical use of ECMO remains limited due to hemocompatibility concerns with pro-coagulatory hollow fiber membrane geometries. Previously, we demonstrated the feasibility of silicon nanopore
Eagleson M.
Concise Encyclopedia Chemistry, 492-492 (1994)
Separation of hexane isomers in a metal-organic framework with triangular channels.
Herm ZR, et al.
Science, 340(6135), 960-964 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service