Skip to Content
Merck
All Photos(3)

Documents

1.00573

Supelco

ortho-Phosphoric acid

85%, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Phosphoric acid, o-Phosphoric acid, Orthophosphoric acid, Orthophosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3PO4
CAS Number:
Molecular Weight:
98.00
Beilstein:
1921286
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21

product name

ortho-Phosphoric acid 85%, for analysis EMSURE® ACS,ISO,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor density

3.4 (vs air)

vapor pressure

2.2 mmHg ( 20 °C)
5 mmHg ( 25 °C)

product line

EMSURE®

form

liquid

concentration

85% phosphoric acid

impurities

≤0.5 ppm As (Arsenic)
≤0.5 ppm Cd (Cadmium)
≤0.5 ppm Co (Cobalt)
≤0.5 ppm Cu (Copper)
≤0.5 ppm Mn (Manganese)
≤0.5 ppm Pb (Lead)
≤1 ppm Fluoride (F)
≤1.0 ppm Ni (Nickel)
≤10 ppm Fe (Iron)
≤10 ppm Heavy metals (as Pb)
≤10 ppm Insoluble matter
≤10 ppm Substances reducing potassium permanganate (as O)
≤10 ppm Volatile acids (as Acetic acid)
≤2 ppm Chloride (Cl)
≤2 ppm Zn (Zinc)
≤20 ppm Ca (Calcium)
≤20 ppm Phosphite and Hypophosphite (as H3PO3)
≤20 ppm Sulfate (SO4)
≤200 ppm Na (Sodium)
≤3 ppm Nitrate (NO3)
≤5 ppm K (Potassium)
≤5 ppm Mg (Magnesium)
≤5 ppm Sb (Antimony)

color

APHA: ≤10

pH

<0.5 (20 °C, 100 g/L in H2O)

kinematic viscosity

30.5 cSt(20 °C)

bp

158 °C (lit.)

mp

~40 °C (lit.)

density

1.685 g/mL at 25 °C (lit.)

storage temp.

no temp limit

SMILES string

OP(O)(O)=O

InChI

1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)

InChI key

NBIIXXVUZAFLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Analysis Note

Assay (alkalimetric)≥ 85.0 %Identitypasses testAppearance of solutionpasses testColor≤ 10 HazenChloride (Cl)≤ 2 ppmFluoride (F)≤ 1 ppmNitrate (NO₃)≤ 3 ppmPhosphite and Hypophosphite (as H₃PO₃)≤ 20 ppmHypophosphite, phosphitepasses testHeavy metals (as Pb)≤ 10 ppmSulfate (SO₄)≤ 20 ppmAs (Arsenic)≤ 0.5 ppmCa (Calcium)≤ 20 ppmCd (Cadmium)≤ 0.5 ppmCo (Cobalt)≤ 0.5 ppmCu (Copper)≤ 0.5 ppmFe (Iron)≤ 10 ppmK (Potassium)≤ 5 ppmMg (Magnesium)≤ 5 ppmMn (Manganese)≤ 0.5 ppmNa (Sodium)≤ 200 ppmNi (Nickel)≤ 1.0 ppmPb (Lead)≤ 0.5 ppmSb (Antimony)≤ 5 ppmZn (Zinc)≤ 2 ppmMatter precipitated by ammoniapasses testVolatile acids (as Acetic acid)≤ 10 ppmSubstances reducing potassium permanganate (as O)≤ 10 ppmInsoluble matter≤ 10 ppm

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

HPLC method on Purospher® RP-18e column analyzes ephedrine and pseudoephedrine HCl in traditional Chinese medicine.

Discovery® C18 column aids in HPLC method development for quantifying ephedrine and pseudoephedrine in traditional Chinese medicine.

Discovery® C18 column aids in HPLC method development for quantifying ephedrine and pseudoephedrine in traditional Chinese medicine.

Discovery® C18 column aids in HPLC method development for quantifying ephedrine and pseudoephedrine in traditional Chinese medicine.

See All

Protocols

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

Sildenafil, sold under the brand name Viagra, among others, is a medication used to treat erectile dysfunction and pulmonary arterial hypertension.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service