Skip to Content
Merck
All Photos(1)

Key Documents

SX0625

Supelco

Sodium Iodide

Meets ACS Specifications GR ACS

Synonym(s):

Sodium Iodide, Sodium Iodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
INa
CAS Number:
Molecular Weight:
149.89
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-679-3
Assay:
≥99.5% (silver nitrate titration)
99.5-100.5% dry basis (silver nitrate titration)
form:
powder
solubility:
1793 g/L

vapor pressure

1 hPa ( 767 °C)

Quality Level

Assay

≥99.5% (silver nitrate titration)
99.5-100.5% dry basis (silver nitrate titration)

form

powder

impurities

≤0.01% Insoluble matter

pH

6-9 (20 °C, 50 g/L in H2O)

bp

1304 °C/1013 hPa

mp

659 °C

solubility

1793 g/L

anion traces

chloride, bromide (as Cl)-: ≤0.01%
iodate (IO3-): ≤0.0003%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.001%
Fe: ≤0.0005%
K: ≤0.01%
Mg: ≤0.001%
heavy metals (as Pb): ≤0.0005%

shipped in

ambient

storage temp.

room temp

SMILES string

[Na+].[I-]

InChI

1S/HI.Na/h1H;/q;+1/p-1

InChI key

FVAUCKIRQBBSSJ-UHFFFAOYSA-M

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay (argentometric)99.5% minBarium (Ba)0.002% maxCalcium (Ca)0.001% maxChloride and bromide (as Cl)0.01% maxHeavy metals (as Pb)0.0005% maxInsoluble matter0.01% maxIodate (IO3)0.0003% maxIron (Fe)0.0005% maxMagnesium (Mg)0.001% maxPhosphate (PO4)0.001% maxPotassium (K)0.01% maxSulfate (SO4)0.002% maxpH (5%, water)6.0-9.0

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service