Skip to Content
Merck
All Photos(2)

Key Documents

32287

Sigma-Aldrich

Heptane

greener alternative

puriss. p.a., reag. Ph. Eur., ≥99% n-heptane basis (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.06
technique(s):
GC/GC: suitable
application(s):
microbiology
bp:
98 °C (lit.)
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

Agency

USP/NF
reag. Ph. Eur.

Quality Level

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

grade

puriss. p.a.

Assay

≥99% n-heptane basis (GC)

form

liquid

autoignition temp.

433 °F

expl. lim.

7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

GC/GC: suitable

impurities

≤0.0005% free acid (as CH3COOH)
≤0.001% non-volatile matter
≤0.005% S-compounds (as S)
≤0.01% water (Karl Fischer)

refractive index

n20/D 1.387 (lit.)
n20/D 1.3870-1.3880

bp

98 °C (lit.)

mp

−91 °C (lit.)

density

0.684 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

suitability

passes test for reaction against H2SO4

SMILES string

CCCCCCC

application(s)

microbiology

greener alternative category

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane and thus has been enhanced for "Safer Solvents and Auxiliaries". Click here for more information.
Heptane (n-Heptane) is a linear hydrocarbon. Its cetane number has been reported to be 56. Mechanism and kinetic studies of its oxidation in flow reactors, shock tubes and rapid compression machines have been proposed.

Application

Greener alternatives to Hexane.
Heptane may be used in the following studies:
  • To compose the water/sodium bis(2-ethylhexyl) sulfosuccinate (AOT)/n-heptane microemulsions, employed for the synthesis of ZnS nanoparticles.
  • As a solvent for various solid n-paraffins.
  • Sterol quantification method (SQM) for the determination of total intracellular ergosterol in Aspergillus fumigatus extracts.

Features and Benefits

Greener alternative for hexane

Other Notes

The article number 32287-4X2.5L will be discontinued. Please order the single bottle 32287-2.5L which is physically identical with the same exact specifications.
The article number 32287-4X2.5L-M will be discontinued. Please order the single bottle 32287-2.5L-M which is physically identical with the same exact specifications.
The article number 32287-6X1L will be discontinued. Please order the single bottle 32287-1L which is physically identical with the same exact specifications.
The article number 32287-6X1L-M will be discontinued. Please order the single bottle 32287-1L-M which is physically identical with the same exact specifications.
Pertains only in Germany: unversteuert

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Márcia Eliana da Silva Ferreira et al.
Antimicrobial agents and chemotherapy, 48(11), 4405-4413 (2004-10-27)
We investigated the evolution of resistance to the antifungal drug itraconazole in replicate populations of Aspergillus fumigatus that were founded from a strain with a genotype of sensitivity to a single drug and then propagated under uniform conditions. For each
Solubility of Long-Chain n-Alkanes in Heptane between 280 and 350 K.
Roberts KL, et al.
Journal of Chemical and Engineering Data, 39(4), 793-795 (1994)
Study of the growth of ZnS nanoparticles in water/AOT/n-heptane microemulsions by UV-absorption spectroscopy.
Calandra P, et al.
Colloids and Surfaces. A, Physicochemical and Engineering Aspects, 160(1), 9-13 (1999)
A comprehensive modeling study of n-heptane oxidation.
Curran HJ, et al.
Combustion and Flame, 114(1), 149-177 (1998)
S Monadjemi et al.
Environmental science & technology, 45(22), 9582-9589 (2011-09-29)
Photodegradation is seldom considered at the surface of vegetation after crop spraying. Chlorothalonil, a broad-spectrum foliar fungicide with a very widespread use worldwide, was considered. To represent the waxy upper layer of leaves, tests were performed within thin paraffin wax

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service