Skip to Content
Merck
All Photos(2)

Key Documents

730491

Sigma-Aldrich

Heptanes, mixture of isomers

greener alternative

contains ≥25% n-heptane, ReagentPlus®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16
CAS Number:
Molecular Weight:
100.20
MDL number:
UNSPSC Code:
12191502
grade:
reagent
technique(s):
HPLC: suitable
bp:
92-100 °C

grade

reagent

product line

ReagentPlus®

Assay

≥99.0%

form

liquid

contains

≥25% n-heptane

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable

impurities

≤0.01% (water)

bp

92-100 °C

density

0.693 g/mL at 25 °C

greener alternative category

SMILES string

C(CCC)CCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent and greener alternative to hexane and thus has been enhanced for "Safer Solvents and Auxiliaries". Click here for more information.

The product is a mixture of heptane isomers with ≥25% n-heptane. The influence of the molecular structure of heptane isomers on the combustion rate was investigated. The thermodynamic changes on mixing heptane isomers with hexadecanes were reported. The performance of Pd/SAPO (palladium/silicoaluminophosphates)-5 and Pd/SAPO-11 molecular seives in the hydroconversion of heptane isomers was studied. Photolysis of heptane isomers by one-color 157nm ionization was analyzed using ion imaging and time-of-flight mass spectroscopy.

Application

Greener alternatives to Hexane in HPLC.

Features and Benefits

Greener alternative for hexane

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroisomerization of heptane isomers over Pd/SAPO molecular sieves: influence of the acid and metal site concentration and the transport properties on the activity and selectivity.
Hochtl M, et al.
J. Catal., 190(2), 419-432 (2000)
Photodissociation of heptane isomers and relative ionization efficiencies of butyl and propyl radicals at 157 nm.
Silva R, et al.
Physical Chemistry Chemical Physics, 11(23), 4777-4781 (2009)
Thermodynamics of mixtures of hexane and heptane isomers with normal and branched hexadecane.
Barbe M and Patterson D.
Journal of Solution Chemistry, 9(10), 753-769 (1980)
The influence of fuel structure on combustion as demonstrated by the isomers of heptane: a rapid compression machine study.
Silke EJ, et al.
Proceedings of the Combustion Institute, 30(2), 2639-2647 (2005)
Heptane as a less toxic option than hexane for the separation of vitamin E from food products using normal phase HPLC.
Buddrick O, et al.
Royal Society of Chemistry Advances, 3, 24063-24068 (2013)

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service