Skip to Content
Merck
All Photos(1)

Key Documents

31665

Sigma-Aldrich

Zinc sulfate heptahydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
powder or crystals

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥99.5%

form

powder or crystals

impurities

≤0.0005% total nitrogen (N)

pH

4.4-5.6 (20 °C, 5%)

anion traces

chloride (Cl-): ≤5 mg/kg

cation traces

As: ≤0.5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤2 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤10 mg/kg
Mg: ≤10 mg/kg
Mn: ≤2 mg/kg
Na: ≤5 mg/kg
Pb: ≤5 mg/kg

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Zinc sulfate heptahydrate has various industrial applications. It is widely employed as an electrolyte for zinc electroplating and for the storage of solar energy in various solar appliances. Its crystals exhibit diamagnetism.

Application

Zinc sulfate heptahydrate may be used in the following studies:
  • To compose electrolyte for the electrodeposition of Cu-Zn-Sn precursors on Mo-coated glass substrates.
  • Preparation of internal standard (IS) working solution for use in liquid chromatography (LC) coupled with mass spectrometry (MS) analysis.
  • Titrimetric estimation of holmium content in the holmium loaded poly(L-lactic acid) films and microspheres.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J F Nijsen et al.
Biomaterials, 22(22), 3073-3081 (2001-09-29)
Holmium-loaded PLLA microspheres are useful systems in radioembolization therapy of liver metastases because of their low density, biodegradability and favourable radiation characteristics. Neutron activated Ho-loaded microspheres showed a surprisingly low release of the relatively small holmium complex. In this paper
Nucleation Kinetics, Growth and Characterization Studies of a Diamagnetic Crystal-Zinc Sulphate Heptahydrate (ZSHH).
Kanagadurai R, et al.
Journal of Chemistry, 6(3), 871-879 (2009)
Organic solvents compromise performance of internal standard (ascomycin) in proficiency testing of mass spectrometry-based assays for tacrolimus.
Kimberly L Napoli
Clinical chemistry, 52(4), 765-766 (2006-04-06)
Preparation of Cu2ZnSnS4 thin films by sulfurization of co-electroplated Cu-Zn-Sn precursors.
Araki H, et al.
Physica Status Solidi, 6(5), 1266-1268 (2009)
Mahmoud Hussein Hadwan et al.
BMC urology, 12, 32-32 (2012-11-14)
Zinc in human seminal plasma is divided into three types of ligands which are high (HMW), intermediate (IMW), and low molecular weight ligands (LMW). The present study was aimed to study the effect of Zn supplementation on the quantitative and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service