Skip to Content
Merck
All Photos(1)

Key Documents

12354

Sigma-Aldrich

Iron(II) sulfate heptahydrate

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-104.5% (manganometric)

Synonym(s):

Ferrous sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeSO4 · 7H2O
CAS Number:
Molecular Weight:
278.01
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

14.6 mmHg ( 25 °C)

grade

puriss.

Assay

99.5-104.5% (manganometric)

form

crystals

quality

meets analytical specification of Ph. Eur., BP, USP, FCC

shelf life

Shelf life is limited

reaction suitability

reagent type: catalyst
core: iron

impurities

organic volatile impurities, complies (GC)
residual solvents, complies
≤0.005% heavy metals (as Pb)

pH

3-4 (20 °C, 5%)

density

1.898 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤100 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤200 mg/kg
Cr: ≤50 mg/kg
Cu: ≤30 mg/kg
Fe3+: ≤2000 mg/kg
Hg: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤200 mg/kg
Mn: ≤1000 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤2 mg/kg
Zn: ≤20 mg/kg

suitability

complies for appearance of solution

SMILES string

O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

InChI

1S/Fe.H2O4S.7H2O/c;1-5(2,3)4;;;;;;;/h;(H2,1,2,3,4);7*1H2/q+2;;;;;;;;/p-2

InChI key

SURQXAFEQWPFPV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Iron(II) sulfate heptahydrate is a heptahydrate of iron(II) sulfate. It undergoes thermal degradation to afford hydrosulfate and oxysulfate of iron(III). Isothermal and dynamic thermal analysis methods have been proposed to study its thermal decomposition in air.

Application

Iron(II) sulfate heptahydrate (FeSO4.7H2O) may be used in the synthesis of maghemite nanoparticles.

Packaging

bottled under inert gas

Caution

tends to oxidation by air

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and characterization of γ-Fe2O3 nanoparticles for applications in magnetic hyperthermia.
Avram A, et al.
Synthesis, 10(P151), 1-1 (2011)
Thermal analysis of iron (II) sulphate heptahydrate in air. V: Thermal decomposition of hydroxy and oxysulphates.
Swamy M and Prasad T.
Journal of Thermal Analysis and Calorimetry, 20(1), 107-114 (1981)
Thermal analysis of iron (II) sulphate heptahydrate in air: III. Thermal decomposition of intermediate hydrates.
Swami MSR and Prasad TP.
Journal of Thermal Analysis and Calorimetry, 19(2), 297-304 (1980)
Yanyan Wei et al.
Food chemistry, 138(2-3), 1952-1958 (2013-02-16)
The present study evaluated the effectiveness of germination and iron fortification on iron concentration and bioavailability of brown rice. Iron fortification during germination process with 0.05-2 g/L ferrous sulfate increased the iron concentration in germinated brown rice from 1.1 to
Hsiao-Chi Chuang et al.
Toxicology and applied pharmacology, 266(3), 329-334 (2012-12-25)
Pulmonary epithelial lining fluid (ELF) is the first substance to make contact with inhaled particulate matter (PM) and interacts chemically with PM components. The objective of this study was to determine the role of ELF in oxidative stress, DNA damage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service