249866
Titanium(IV) chloride solution
1.0 M in methylene chloride
Synonym(s):
Titanium tetrachloride
About This Item
Recommended Products
grade
for analytical purposes
Quality Level
vapor pressure
24.09 psi ( 55 °C)
6.93 psi ( 20 °C)
form
liquid
reaction suitability
reagent type: catalyst
core: titanium
concentration
0.95-1.10 M NaOH (titration)
1.0 M in methylene chloride
density
1.365 g/mL at 25 °C
SMILES string
Cl[Ti](Cl)(Cl)Cl
InChI
1S/4ClH.Ti/h4*1H;/q;;;;+4/p-4
InChI key
XJDNKRIXUMDJCW-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service