Skip to Content
Merck
All Photos(1)

Key Documents

238937

Sigma-Aldrich

Drierite

without indicator, 8 mesh

Synonym(s):

Calcium sulfate, Gypsum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4
CAS Number:
Molecular Weight:
136.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NB.24
Pricing and availability is not currently available.

Quality Level

form

chips

particle size

8 mesh

SMILES string

[Ca++].[O-]S([O-])(=O)=O

InChI

1S/Ca.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

OSGAYBCDTDRGGQ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Drierite can be used as a desiccant to dry solvents in various reactions. For example, it is used to dry acetone in the preparation of C-H activated Ir(I) acetonyl complex[1].

Legal Information

A product of Reilly Industries, Inc. ®W.A. Hammond Drierite Co.
Drierite is a trademark of W.A. Hammond Drierite Co., LTD

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iridium (i) PNP complexes in the sp 3 C--H bond activation of methyl propanoate and related esters
Coetzee J, et al.
Dalton Transactions, 44, 1585-1591 (2015)
GAS CHROMATOGRAPHIC DETERMINATION OF LOWER ALCOHOLS IN BIOLOGIC SAMPLES.
H LYONS et al.
Clinical chemistry, 10, 429-432 (1964-05-01)
The ontogeny of personality in the wild guinea pig.
Guenther A, et al.
Animal behaviour, 90, 131-139 (2014)
Respirometry: anhydrous drierite equilibrates with carbon dioxide and increases washout times.
White CR, et al.
Physiological and Biochemical Zoology : Pbz, 79(5), 977-988 (2006)
The analysis of carbon monoxide in urban air at the ppm level, and the normal carbon monoxide value.
L Dubois et al.
Journal of the Air Pollution Control Association, 16(3), 135-139 (1966-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service