Skip to Content
Merck
All Photos(1)

Documents

12090

Sigma-Aldrich

Calcium sulfate hemihydrate

purum, ≥97.0%

Synonym(s):

Calcium sulphate hemihydrate, Gypsum hemihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaSO4 · 0.5H2O
CAS Number:
Molecular Weight:
145.15
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

purum

Quality Level

Assay

≥97.0%

SMILES string

O.[Ca++].[Ca++].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/2Ca.2H2O4S.H2O/c;;2*1-5(2,3)4;/h;;2*(H2,1,2,3,4);1H2/q2*+2;;;/p-4

InChI key

ZOMBKNNSYQHRCA-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Angela R Roper et al.
Health physics, 104(3), 264-269 (2013-01-31)
Coal combustion residues from coal-fired power plants can be advantageous for use in building and construction materials. These by-products contain trace quantities of naturally occurring radionuclides from the uranium and thorium series, as well as other naturally occurring radionuclides such
Clovis Monteiro Bramante et al.
Journal of applied oral science : revista FOB, 21(1), 32-36 (2013-04-06)
To evaluate the biocompatibility and the setting time of Portland cement clinker with or without 2% or 5% calcium sulfate and MTA-CPM. Twenty-four mice (Rattus norvegicus) received subcutaneously polyethylene tubes filled with Portland cement clinker with or without 2% or
Simple design for an aeroplane plaster of Paris splint in the immobilisation of axillary skin grafts.
A Salibi et al.
Annals of the Royal College of Surgeons of England, 95(4), 298-298 (2013-05-17)
Massimo Peruffo et al.
Physical chemistry chemical physics : PCCP, 15(6), 1956-1965 (2012-12-22)
Dissolution processes at single crystal surfaces often involve the initial formation and expansion of localized, characteristic (faceted) etch-pits at defects, in an otherwise comparatively unreactive surface. Using natural gypsum single crystal as an example, a simple but powerful morphological analysis
Sung-Yen Lin et al.
The Journal of hand surgery, 38(1), 104-109 (2012-12-04)
Enchondroma of the hand with a pathologic fracture is generally treated by tumor curettage and bone grafting after the fracture has healed. However, delayed surgery postpones definitive diagnosis and prolongs the period of disability. We have treated pathologic fractures in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service