Skip to Content
Merck
All Photos(3)

Key Documents

224820

Sigma-Aldrich

Ammonium hydrogen difluoride

reagent grade, 95%

Synonym(s):

Ammonium bifluoride, Etching powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4HF2
CAS Number:
Molecular Weight:
57.04
EC Number:
MDL number:
UNSPSC Code:
12352300
eCl@ss:
38050109
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
crystals
flakes

grade

reagent grade

Assay

95%

form

crystals
flakes

mp

125 °C (lit.)

SMILES string

N.F.F

InChI

1S/2FH.H3N/h2*1H;1H3

InChI key

KVBCYCWRDBDGBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Employed in a new hydrothermal synthesis of complex flourides NaHoF4 and NaEuF4 these interesting compounds have the fluorite structure and are important forsolid-state lasers as well as for photoluminescence and magnetic characteristics.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xun, X. et al.
Chemistry of Materials, 9, 2966-2966 (1997)
Na-Yu Chia et al.
Gut, 64(5), 707-719 (2014-07-24)
Gastric cancer (GC) is a deadly malignancy for which new therapeutic strategies are needed. Three transcription factors, KLF5, GATA4 and GATA6, have been previously reported to exhibit genomic amplification in GC. We sought to validate these findings, investigate how these
Kevin R Bushell et al.
Blood, 125(6), 999-1005 (2014-12-04)
Non-Hodgkin lymphomas (NHLs) are the most common cancer to affect pet dogs. In contrast to the many genes whose mutation contributes to lymphomagenesis in humans, relatively little is known about the acquired genetic alterations that lead to canine B-cell lymphomas
Bishi Fu et al.
Journal of virology, 89(1), 195-207 (2014-10-17)
We have previously shown that ORF45, an immediate-early and tegument protein of Kaposi's sarcoma-associated herpesvirus (KSHV), causes sustained activation of p90 ribosomal S6 kinases (RSKs) and extracellular regulated kinase (ERK) (E. Kuang, Q. Tang, G. G. Maul, and F. Zhu
Ryo Jimbo et al.
Clinical implant dentistry and related research, 10(1), 55-61 (2008-02-08)
Previously, we reported that anodized porous titanium implants have photocatalytic hydrophilicity. However, this effect was not always sufficient for the significant improvement of bone apposition. The purpose of this study was to improve the photocatalytic properties of porous titanium implants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service