Skip to Content
Merck
Ammonium hydrogen difluoride SAJ special grade, ≥98.5%

Ammonium hydrogen difluoride

Ammonium hydrogen difluoride SAJ special grade, ≥98.5%
Synonym(s):
Ammonium bifluoride, Etching powder
Linear Formula:
NH4HF2
CAS Number:
Molecular Weight:
57.04
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment