Skip to Content
Merck
All Photos(3)

Documents

199443

Sigma-Aldrich

Aluminum oxide

activated, basic, Brockmann I

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
SB.52

grade

Brockmann I
activated
standard grade

Quality Level

Agency

suitable for EPA 1613

form

powder

manufacturer/tradename

CAMAG 5016-A-I

technique(s)

solid phase extraction (SPE): suitable

impurities

<0.03% Fe2O3
<0.4% Na2O

particle size

~150 mesh

pore size

58 Å pore size

pH

9.5±0.5 ( in H2O)

mp

2040 °C (lit.)

anion traces

chloride (Cl-): none detected
silicate (as SiO2): <0.03%

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration.
The product is basic activated aluminum oxide (Alumina, Al2O3) having an activity grade Brockman I. The mechanism of formation of protective scales of α-Al2O3 on surfaces of binary M-Al (M = Fe, Ni or Co) alloys at high temperatures has been studied. Activated aluminum oxides having different acidities and water concentrations (having Brockmann activity I-V) can easily detect various types of water (namely free water, pore water, and bound water) on the aluminum oxide present in soils.

Application

Aluminum oxide (Alumina) has been used for the removal of peroxides from the dehydration solvent (tetrahydrofuran, THF) in a protocol. Protocol involved the high resolution study of fine brain anatomy of fluorescently labeled mouse brains.
It may be used in the fabrication of polycrystalline lithium iodide-aluminum oxide solid electrolytes. The incorporation of aluminum oxide remarkably increased the conductivity of lithium iodide.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micron-scale resolution optical tomography of entire mouse brains with confocal light sheet microscopy.
Silvestri L, et al.
Journal of Visualized Experiments, 80, e50696-e50696 (2013)
Conduction Characteristics of the Lithium Iodide-Aluminum Oxide Solid Electrolytes.
Liang CC.
Journal of the Electrochemical Society, 120(10), 1289-1292 (1973)
Prescott R and Graham MJ.
Oxidation of Metals, 38(3-4), 233-254 (1992)
Thermoanalytical studies of water on activated alumina, Brockmann I-V,(acid, neutral, basic) from- 60? to+ 700? C.
Hampson JW and Bleam WF.
Thermochimica Acta, 288(1), 179-189 (1996)
Colin J Ingham et al.
Biotechnology advances, 30(5), 1089-1099 (2011-08-23)
Porous aluminum oxide (PAO) is a ceramic formed by an anodization process of pure aluminum that enables the controllable assembly of exceptionally dense and regular nanopores in a planar membrane. As a consequence, PAO has a high porosity, nanopores with

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service