Skip to Content
Merck
All Photos(1)

Key Documents

06300

Supelco

Aluminum oxide Inorganic Sorbent

activated, neutral, 100-285 mesh, Brockmann Activity I

Synonym(s):

Alumina

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al2O3
CAS Number:
Molecular Weight:
101.96
EC Number:
MDL number:
UNSPSC Code:
12352311
eCl@ss:
38120402
PubChem Substance ID:
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

Aluminum oxide, activated, neutral, Brockmann Activity I

grade

standard

Quality Level

form

powder

quality

activated, neutral

technique(s)

LPLC: suitable

surface area

155 m2/g

matrix

Aluminum oxide

matrix active group

aluminum oxide

particle size

0.05-0.15 mm

pore size

60 Å mean pore size

pH

7.0±0.5

mp

2040 °C (lit.)

separation technique

hydrophilic interaction (HILIC)

SMILES string

O=[Al]O[Al]=O

InChI

1S/2Al.3O

InChI key

TWNQGVIAIRXVLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Brockmann aluminum oxides are suitable for column chromatography and adsorptive filtration.
Aluminum oxides are generally used as solid support phases in solid phase extraction (SPE)-based clean-up procedures employed in chromatographic methods.[1]

Application

Aluminum oxide has been used as a chromatographic column support for the purification of sn-2 monopalmitin prior to its determination by high performance liquid chromatography-evaporative light scattering detection (HPLC-ELSD).[2]
It may be used for purification of fish tissue extract followed by liquid−liquid extraction into dichloromethane prior to the analysis for triarylmethane and phenothiazine dyes by liquid chromatography-tandem mass spectrometry (LC−MS/MS).[1] It may also be used as a solid support phase for SPE-based analysis of pyrethroid insecticide residues in vegetable oils by gas chromatography-tandem mass spectrometry (GC-MS/MS).[3]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiresidue determination of triarylmethane and phenothiazine dyes in fish tissues by LC-MS/MS.
Tarbin JA, et al.
Analytica Chimica Acta, 625(2), 188-194 (2008)
Determination of pyrethroid insecticide residues in vegetable oils by using combined solid-phases extraction and tandem mass spectrometry detection.
Esteve-Turrillas FA, et al.
Analytica Chimica Acta, 553(1-2), 50-57 (2005)
A López-López et al.
Journal of chromatography. B, Biomedical sciences and applications, 760(1), 97-105 (2001-08-28)
An alternative method to determine the sn-2 monopalmitin in infant formulas was developed and validated. This method offers many advantages over the traditional methods. It follows the official method in the first steps, purification of the fat or oil through
Chien-Chih Lin et al.
Nanoscale, 5(17), 8090-8097 (2013-07-25)
We demonstrated a promising route for enhancing temperature sensitivity, improving saturation voltage, and reducing power consumption of the MOS(p) tunneling temperature sensors by introducing ultrathin Al2O3 into the dielectric stacks. Detailed illustrations of the working mechanism and device concept are
Jaakko Akola et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(25), 10129-10134 (2013-06-01)
Glass formation in the CaO-Al2O3 system represents an important phenomenon because it does not contain typical network-forming cations. We have produced structural models of CaO-Al2O3 glasses using combined density functional theory-reverse Monte Carlo simulations and obtained structures that reproduce experiments

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service