Skip to Content
Merck
All Photos(5)

Key Documents

155810

Sigma-Aldrich

2-Methyltetrahydrofuran

greener alternative

BioRenewable, ReagentPlus®, ≥99.5%, contains 150-400 ppm BHT as stabilizer

Synonym(s):

2-MeTHF, Tetrahydro-2-methylfuran, Tetrahydrosilvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102448
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

≥99.5%

form

liquid

contains

150-400 ppm BHT as stabilizer

expl. lim.

0.34-6.3 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1.0% water (coulometr.)
≤100 ppm peroxide (as H2O2)

refractive index

n20/D 1.406 (lit.)

bp

78-80 °C (lit.)

mp

-136 °C

density

0.86 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

CC1CCCO1

InChI

1S/C5H10O/c1-5-3-2-4-6-5/h5H,2-4H2,1H3

InChI key

JWUJQDFVADABEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product is biorenewable and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks".  Click here  for more information.

2-Methyltetrahydrofuran (2-MTHF), a 2-methyl substituted tetrahydrofuran, is a biomass derived, environmentally favorable solvent alternative to tetrahydrofuran (THF), 1,4 Dioxane (Dioxane) and dichloromethane (DCM) for most industrial applications. Its polarity and Lewis base strength is intermediate between tetrahydrofuran (THF) and diethyl ether. It is a potential greener solvent alternative for organic synthesis and shows resistance to reduction by lithium making it a promising candidate as electrolytes in lithium batteries.

Application

2-Methyltetrahydrofuran may be used as an alternative solvent to:
THF in organometallic reactions
  • Grignard
  • Reformantsky
  • Lithiation
  • Hydride Reduction
  • Metal-Catalyzed Coupling (Heck, Stile, Suzuki)




Dichloromethane for Biphasic Reactions:
  • Alkylation
  • Amidation
  • Nucleophilic Substitution

Features and Benefits

  • Made from Renewable Resource - furfural derived from agricultural byproducts
  • High boiling point
  • Clean organic-water phase separation
  • Reduced energy to recover

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mari Merce Cascant et al.
Analytical and bioanalytical chemistry, 409(14), 3527-3539 (2017-04-09)
There is a great interest in finding alternatives and green solvents in extraction processes to replace petroleum based solvents. In order to investigate these possibilities, computational methods, as Hansen solubility parameters (HSP) and conductor-like screening model for real solvent (COSMO-RS)
V R Koch et al.
Science (New York, N.Y.), 204(4392), 499-501 (1979-05-04)
Rechargeable, high energy density lithium batteries require an efficient lithium electrode. Earlier work with electrolytes based on propylene carbonate, methyl acetate, and tetrahydrofuran yielded poor lithium electrode cycling efficiencies because of electrolyte reduction by lithium. Solutions of lithium hexafluoroarsenate in
Solvent applications of 2-methyltetrahydrofuran in organometallic and biphasic reactions.
Aycock DF.
Organic Process Research & Development, 11(1), 156-159 (2007)
Axel Kahnt et al.
Journal of the American Chemical Society, 133(25), 9863-9871 (2011-05-21)
Electron-transfer reactions are fundamental to many practical devices, but because of their complexity, it is often very difficult to interpret measurements done on the complete device. Therefore, studies of model systems are crucial. Here the rates of charge separation and
Tony W T Bristow et al.
Journal of the American Society for Mass Spectrometry, 25(10), 1794-1802 (2014-08-12)
For on-line monitoring of chemical reactions (batch or continuous flow), mass spectrometry (MS) can provide data to (1) determine the fate of starting materials and reagents, (2) confirm the presence of the desired product, (3) identify intermediates and impurities, (4)

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service