Skip to Content
Merck
All Photos(1)

Key Documents

13464

Sigma-Aldrich

Sodium sulfate

puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO4
CAS Number:
Molecular Weight:
142.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystalline

grade

puriss.

Assay

99.0-100.5% (calc. to the dried substance)

form

crystalline

quality

meets analytical specification of Ph. Eur., BP, USP

impurities

acidity or alkalinity, complies
residual solvents, complies
≤0.001% heavy metals (as Pb)
≤0.04% free alkali (as NaOH)
≤0.05% free acid (as H2SO4)

loss

≤0.5% loss on drying, 130 °C

pH

5.2-8 (20 °C, 50 g/L)

mp

884 °C (lit.)

density

2.68 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤200 mg/kg

cation traces

As: ≤2 mg/kg
Ca: ≤50 mg/kg
Fe: ≤50 mg/kg
Mg: ≤50 mg/kg
Zn: ≤300 mg/kg

suitability

complies for appearance of solution

SMILES string

[Na+].[Na+].[O-]S([O-])(=O)=O

InChI

1S/2Na.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

PMZURENOXWZQFD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfate (Na2SO4) can be obtained directly via precipitation from a saturated solution of Na2SO4 at room temperature (20°C). Its crystallization and growth rates at room temperature (20°C) under various conditions have been investigated. Sodium sulfate can be produced during Leblanc process, from sodium chloride and sulfuric acid.

Application

Sodium sulfate (Na2SO4) may be employed as a reagent for the precipitation of globulins during the estimation of proteins in blood.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas M
Sodium (2004)
The use of sodium sulfate as the globulin precipitant in the determination of proteins in blood.
Howe PE.
The Journal of Biological Chemistry, 49(1), 93-107 (1921)
How does sodium sulfate crystallize? Implications for the decay and testing of building materials.
Rodriguez-Navarro C, et al.
Cement and Concrete Research, 30(10), 1527-1534 (2000)
Teppei Adachi et al.
Inflammatory bowel diseases, 21(1), 31-39 (2014-12-02)
Expression of heat shock protein A4 (HSPA4, also called Apg-2), a member of the HSP110 family, is induced by several forms of stress. The physiological and pathological functions of HSPA4 in the intestine remain to be elucidated. We assessed HSPA4
Clément Cartier et al.
Water research, 47(2), 661-671 (2012-11-24)
Release of lead from 80% partially replaced service lines was compared to full lead service lines using harvested-stabilized lead pipes and field brass connectors. After more than a year of stabilization, lead release was consistent with field samples. Over the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service