Skip to Content
Merck
All Photos(7)

Key Documents

1.00003

Supelco

Acetonitrile

≥99.5% (GC), ACS reagent, for extraction and analytical purposes, Reag. Ph Eur, EMSURE®

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-835-2
NACRES:
NB.21

product name

Acetonitrile, for analysis EMSURE® ACS,Reag. Ph Eur

grade

ACS reagent
for analytical purposes
for extraction
reagent grade

Quality Level

Agency

reag. Ph. Eur.

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)
97 hPa ( 20 °C)

product line

EMSURE®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

524 °C
973 °F

expl. lim.

16 %

impurities

≤0.0001 meq/g Alkalinity
≤0.0002 meq/g Acidity
≤0.005% Cyanide (CN)
≤0.1% Water

evapn. residue

≤0.001%

color

clearAPHA: ≤10

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

transition temp

flash point 2 °C

density

0.786 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.00005%
B: ≤0.000002%
Ba: ≤0.00001%
Ca: ≤0.00005%
Cd: ≤0.000005%
Co: ≤0.000002%
Cr: ≤0.000002%
Cu: ≤0.000002%
Fe: ≤0.00001%
Mg: ≤0.00001%
Mn: ≤0.000002%
Ni: ≤0.000002%
Pb: ≤0.00001%
Sn: ≤0.00001%
Zn: ≤0.00001%

SMILES string

CC#N

application(s)

environmental
food and beverages
industrial qc
pharmaceutical
sample preparation

format

neat

storage temp.

2-30°C

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.

Application


  • Modified QuEChERS method combined with ultra-performance liquid chromatography-tandem mass spectrometry: This research utilizes acetonitrile′s excellent solvent properties for the detection of cyclopiazonic acid in feeds. The method enhances the efficiency and accuracy of chemical analyses in food safety, demonstrating acetonitrile′s pivotal role in ensuring the quality of agricultural products (Peng et al., 2024).

  • Liquid Chromatographic Enantioseparation of Newly Synthesized Fluorinated Tryptophan Analogs: Highlighting its use in pharmaceutical research, acetonitrile is employed in the chromatographic enantioseparation of complex amino acids, essential for drug development and molecular pharmacology studies (Tanács et al., 2024).

  • Synthesis and Application of 1,8-Naphthalimide Derivatives Fluorescent Probe: Acetonitrile′s versatility is showcased in the synthesis of fluorescent probes for environmental monitoring. Its application in detecting and measuring environmental pollutants highlights its significant impact on ecological research and safety (Negi et al., 2024).

Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0001 meq/g
Density (d 20 °C/20 °C): 0.782 - 0.783
Refractive index (n 20/D): 1.343 - 1.345
Boiling range (80-82°C): ≥ 95 % (v/v)
Cyanide (CN): ≤ 0.005 %
Al (Aluminium): ≤ 0.00005 %
B (Boron): ≤ 0.000002 %
Ba (Barium): ≤ 0.00001 %
Ca (Calcium): ≤ 0.00005 %
Cd (Cadmium): ≤ 0.000005 %
Co (Cobalt): ≤ 0.000002 %
Cr (Chromium): ≤ 0.000002 %
Cu (Copper): ≤ 0.000002 %
Fe (Iron): ≤ 0.00001 %
Mg (Magnesium): ≤ 0.00001 %
Mn (Manganese): ≤ 0.000002 %
Ni (Nickel): ≤ 0.000002 %
Pb (Lead): ≤ 0.00001 %
Sn (Tin): ≤ 0.00001 %
Zn (Zinc): ≤ 0.00001 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.1 %
ACS, Ph Eur-reagent

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

HPLC method determines Drospirenone in progestin medication using Purospher® STAR RP-18e Hibar® HPLC column, meeting USP monograph criteria.

HPLC method determines Drospirenone in progestin medication using Purospher® STAR RP-18e Hibar® HPLC column, meeting USP monograph criteria.

HPLC method determines Drospirenone in progestin medication using Purospher® STAR RP-18e Hibar® HPLC column, meeting USP monograph criteria.

HPLC method determines Drospirenone in progestin medication using Purospher® STAR RP-18e Hibar® HPLC column, meeting USP monograph criteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service