Skip to Content
Merck
All Photos(1)

Key Documents

10229

Sigma-Aldrich

Silver sulfate

puriss., ≥99%

Synonym(s):

Sulfuric acid disilver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2SO4
CAS Number:
Molecular Weight:
311.80
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99%
form:
powder or crystals
Pricing and availability is not currently available.

grade

puriss.

Quality Level

Assay

≥99%

form

powder or crystals

reaction suitability

reagent type: catalyst
core: silver

mp

652 °C (lit.)

anion traces

chloride (Cl-): ≤10 mg/kg
nitrate (NO3-): ≤100 mg/kg

cation traces

Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
Pb: ≤50 mg/kg

SMILES string

[Ag+].[Ag+].[O-]S([O-])(=O)=O

InChI

1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

YPNVIBVEFVRZPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Silver sulfate (Ag2SO4) is the disilver(I) salt of sulfuric acid. It participates in the TiO2-assisted photooxidation of substituted benzyltrimethylsilanes in acetonitrile. Diarylethanes were obtained in 50-74% yields.[1] It has been reported as one of the common atmospheric silver corrosion product.[2]

Application

Silver sulfate may be employed in the following studies:
  • Iodination reagent in combination with iodine for the synthesis of iododerivatives.[3]
  • Synthesis of iodinated uredines.[4]
  • Preparation of silver sulfate reagent, which is required for the determination of Chemical Oxygen Demand (COD).[5]

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical stability of plasmon-active silver tips for tip-enhanced Raman spectroscopy.
Kalbacova J, et al.
Nanospectroscopy, 1(1), 12-18 (2014)
Ponnusamy Velusamy et al.
Journal of advanced research, 5(1), 19-25 (2015-02-17)
The photocatalytic decoloration of an organic dye, ethyl violet (EV), has been studied in the presence of TiO2 and the addition of β-Cyclodextrin (β-CD) with TiO2 (TiO2-β-CD) under UV-A light irradiation. The different operating parameters like initial concentration of dye
Iodination with Silver Sulfate and Iodine. II. Uridines.
Sy W-W.
Synthetic Communications, 20(21), 3391-3394 (1990)
Iodination of aromatic amines with iodine and silver sulfate.
Sy W-W.
Synthetic Communications, 22(22), 3215-3219 (1992)
Titanium dioxide photocatalysed oxidation of benzyltrimethylsilanes in the presence of silver sulfate.
Baciocchi E, et al.
Journal of the Chemical Society. Chemical Communications, 1, 59-60 (1992)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service