Skip to Content
Merck
All Photos(1)

Key Documents

10029

Sigma-Aldrich

Mercury(II) sulfate

puriss., ≥98%

Synonym(s):

Mercuric sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HgSO4
CAS Number:
Molecular Weight:
296.65
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
form:
powder or crystals

grade

puriss.

Assay

≥98%

form

powder or crystals

ign. residue (after red. with HCOOH)

≤0.1% (as SO4)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

[Hg++].[O-]S([O-])(=O)=O

InChI

1S/Hg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

DOBUSJIVSSJEDA-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Mercury(II) sulfate can be used as a catalyst in the presence of sulfuric acid (H2SO4) for the:
  • Direct oxidation of methane to methanol.
  • Hydration of alkynes to the corresponding carbonyl compounds.
  • Conversion of alkyl protected glycals into their corresponding α,β-unsaturated enals (Perlin aldehydes).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laurie F Caslake et al.
Applied and environmental microbiology, 70(2), 1145-1150 (2004-02-10)
Contaminated water causes an estimated 6 to 60 billion cases of gastrointestinal illness annually. The majority of these cases occur in rural areas of developing nations where the water supply remains polluted and adequate sanitation is unavailable. A portable, low-cost
Direct oxidation of methane to methanol by mercuric sulfate catalyst.
Gang X, et al.
J. Catal., 196(2), 287-292 (2000)
Tariq SA and Hill JO.
J. Therm. Anal., 212, 277-281 (1981)
Catalysis by mercuric ion of reactions of glycals with water.
Gonzalez F, et al.
Carbohydrate Research, 42(2), 267-274 (1975)
A P de Souza et al.
Dental materials : official publication of the Academy of Dental Materials, 16(2), 103-108 (2001-02-24)
The interaction between metal ions and the oral environment is a major subject matter in dental research. Matrix metalloproteinases (MMPs) have been implicated in several pathologic oral processes such as periodontal tissue destruction, root caries, tumour invasion and temporomandibular joint

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service