Skip to Content
Merck
All Photos(1)

Key Documents

Y0001074

Rifaximin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Deoxy-4′-methylpyrido[1′,2′-1,2]imidazo[5,4-c]rifamycin SV, Rifacol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C43H51N3O11
CAS Number:
Molecular Weight:
785.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rifaximin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)C(C)=C\C=C\[C@@H](C)[C@@H](O)[C@@H](C)[C@H](O)[C@H](C)[C@H]([C@H]1C)C(=O)OC)c5c(nc6cc(C)ccn56)c4c3C2=O

InChI

1S/C43H51N3O11/c1-19-14-16-46-27(18-19)44-32-29-30-37(49)25(7)39-31(29)40(51)43(8,57-39)56-17-15-26(54-9)22(4)28(42(53)55-10)23(5)36(48)24(6)35(47)20(2)12-11-13-21(3)41(52)45-33(34(32)46)38(30)50/h11-18,20,22-24,26,28,35-36,47-50H,1-10H3,(H,45,52)/b12-11+,17-15+,21-13-/t20-,22+,23-,24-,26+,28+,35-,36-,43+/m1/s1

InChI key

HIYLTQREEOINNF-HTEWPBCCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifaximin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Belous et al.
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology, (3)(3), 63-68 (2013-12-04)
One of the important problems in the treatment of patients with inflammatory bowel disease are the persistent complaints of pain, abdominal distention, frequent stools, excretion of mucus with faeces with the presence of endoscopic or roentgenologic remission in the damaged
Rima I El-Herte et al.
Scandinavian journal of infectious diseases, 44(3), 228-230 (2011-11-15)
Clostridium difficile colitis infection is on the rise and is considerably increasing the duration of hospital stay, as well as healthcare costs. The management of C. difficile colitis has become more challenging with the increasing failure of therapeutic response to
Anastasia Rivkin et al.
Clinical therapeutics, 33(7), 812-827 (2011-07-12)
Rifaximin is a nonabsorbable oral antibiotic that acts locally in the gastrointestinal tract with minimal systemic adverse effects. Rifaximin received new labeling for reduction in the risk of the recurrence of overt hepatic encephalopathy (HE) in patients with advanced liver
David J Farrell
Journal of clinical gastroenterology, 47(3), 205-211 (2013-01-24)
Irritable bowel syndrome (IBS), a chronic, nonfatal illness is commonly encountered in clinical practice; however, treatment options are limited and often ineffectual. Despite this, there is increasing evidence that bacterial overgrowth in the bowel (dysbiosis) may be an etiological factor
Kenneth R Lawrence et al.
Pharmacotherapy, 28(8), 1019-1032 (2008-07-29)
To review the effectiveness and safety of rifaximin for the treatment of hepatic encephalopathy. A literature search was conducted of MEDLINE (1966-September 2007), the Cochrane Database of Systematic Reviews (1995-2007), and the Cochrane Hepato-Biliary Group Reviews (2003-2007). English-language articles identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service