Skip to Content
Merck
All Photos(1)

Key Documents

R0900000

Rifamycin B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Rifamycin impurity A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H49NO14
CAS Number:
Molecular Weight:
755.80
Beilstein:
604953
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

rifamycin, rifampicin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C39H49NO14/c1-17-11-10-12-18(2)38(49)40-24-15-26(51-16-27(42)43)28-29(34(24)47)33(46)22(6)36-30(28)37(48)39(8,54-36)52-14-13-25(50-9)19(3)35(53-23(7)41)21(5)32(45)20(4)31(17)44/h10-15,17,19-21,25,31-32,35,44-47H,16H2,1-9H3,(H,40,49)(H,42,43)/b11-10+,14-13+,18-12-/t17-,19+,20+,21+,25-,31-,32+,35+,39-/m0/s1

InChI key

SQTCRTQCPJICLD-KTQDUKAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Rifamycin B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H G Floss
Journal of industrial microbiology & biotechnology, 27(3), 183-194 (2002-01-10)
The evolution of the field of biosynthesis from the unravelling of the mode of formation of natural products to the use of such knowledge to create new compounds is reviewed using examples from the author's laboratory. The discussion focuses on
Prashant M Bapat et al.
Biotechnology and bioengineering, 93(4), 779-790 (2005-11-23)
Industrial fermentations typically use media that are balanced with multiple substitutable substrates including complex carbon and nitrogen source. Yet, much of the modeling effort to date has mainly focused on defined media. Here, we present a structured model that accounts
A Courtois et al.
Cancer letters, 139(1), 97-104 (1999-07-17)
The multidrug resistance-associated protein (MRP) is a drug efflux membrane pump conferring multidrug resistance on tumor cells. In order to look for compounds that can lead to reversal of such a resistance, the antituberculosis compound rifampicin, belonging to the chemical
Ansgar Stratmann et al.
The Journal of antibiotics, 55(4), 396-406 (2002-06-14)
Proansamycin B, the formerly postulated intermediate of rifamycin B biosynthesis, was isolated from cultures of the Amycolatopsis mediterranei mutant F1/24. The structure was determined using UV, IR, NMR and MS techniques. Biotransformation studies demonstrate that proansamycin B is an intermediate
Zhi-Hua Jin et al.
Journal of Zhejiang University. Science, 5(12), 1590-1596 (2004-11-18)
Study of the effect of dissolved oxygen and shear stress on rifamycin B fermentation with A. mediterranei XC 9-25 showed that rifamycin B fermentation with Amycolatoposis mediterranei XC 9-25 needs high dissolved oxygen and is not very sensitive to shearing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service