Skip to Content
Merck
All Photos(1)

Key Documents

Y0000647

Benzophenone

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Diphenyl ketone, Diphenylmethanone, NSC 8077

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
Beilstein:
1238185
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

Agency

EP Reference Standard

vapor pressure

1 mmHg ( 108 °C)

API family

benzophenone

manufacturer/tradename

EDQM

bp

305 °C (lit.)

mp

47-51 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzophenone EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Pockrandt et al.
Chembiochem : a European journal of chemical biology, 13(18), 2764-2771 (2012-11-15)
Gene-inactivation experiments have indicated that the putative prenyltransferase XptB from Aspergillus nidulans was likely to be responsible for the prenylation of 1,7-dihydroxy-6-methyl-8-hydroxymethylxanthone. Recently, it was suggested that this enzyme might also accept as substrate the benzophenone arugosin H, which is
Masayuki Kyomoto et al.
Biomaterials, 34(32), 7829-7839 (2013-07-31)
We investigated the production of free radicals on a poly(ether-ether-ketone) (PEEK) substrate under ultraviolet (UV) irradiation. The amount of the ketyl radicals produced from the benzophenone (BP) units in the PEEK molecular structure initially increased rapidly and then became almost
Young-Jin Kim et al.
Angewandte Chemie (International ed. in English), 51(42), 10537-10541 (2012-09-20)
Caught in a web: Photo-cross-linkable temperature-responsive polymer-based nanofiber webs have been synthesized that have the ability to capture, encapsulate, and release cells by dynamically transforming the fibrous structure into hydrogel-like structures by wrapping, swelling, and deswelling processes in response to
Ashna A Khan et al.
Organic & biomolecular chemistry, 11(6), 881-885 (2012-12-22)
Trehalose dimycolates (TDMs) are the most abundant glycolipids found in the cell wall of Mycobacterium tuberculosis (M. tb). TDMs play an important role in the pathogenesis of M. tb yet the only known receptor for TDM is the macrophage inducible
Tao Zhang et al.
The Science of the total environment, 461-462, 49-55 (2013-05-29)
Limited information exists on the exposure of benzophenone (BP)-type UV filters (i.e., sunscreen compounds) in children, adults, and pregnant women in China. In this study, we determined the concentrations of five BP derivatives, BP-1, BP-2, BP-3, BP-8, and 4OH-BP in

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service