Merck
A smart nanofiber web that captures and releases cells.
Angewandte Chemie (International ed. in English) (2012-09-20)
Young-Jin Kim, Mitsuhiro Ebara, Takao Aoyagi
ABSTRACT
Caught in a web: Photo-cross-linkable temperature-responsive polymer-based nanofiber webs have been synthesized that have the ability to capture, encapsulate, and release cells by dynamically transforming the fibrous structure into hydrogel-like structures by wrapping, swelling, and deswelling processes in response to external temperature changes (see picture).
Product Description
Supelco
Melting point standard 47-49°C, analytical standard
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Supelco
Mettler-Toledo Calibration substance ME 18870, Benzophenone, traceable to primary standards (LGC)
Sigma-Aldrich
Benzophenone, SAJ first grade, ≥99.0%
Supelco
Benzophenone, analytical standard
Sigma-Aldrich
Benzophenone, purified by sublimation, ≥99%
Sigma-Aldrich
Benzophenone, ReagentPlus®, 99%
Supelco
Benzophenone, Pharmaceutical Secondary Standard; Certified Reference Material
Benzophenone, European Pharmacopoeia (EP) Reference Standard