Skip to Content
Merck
All Photos(1)

Key Documents

Y0000517

Glutathione

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

L-Glutathione reduced, γ-L-Glutamyl-L-cysteinyl-glycine, GSH

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH(CO2H)CH2CH2CONHCH(CH2SH)CONHCH2CO2H
CAS Number:
Molecular Weight:
307.32
Beilstein:
1729812
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

glutathione

manufacturer/tradename

EDQM

mp

192-195 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](CCC(=O)N[C@@H](CS)C(=O)NCC(O)=O)C(O)=O

InChI

1S/C10H17N3O6S/c11-5(10(18)19)1-2-7(14)13-6(4-20)9(17)12-3-8(15)16/h5-6,20H,1-4,11H2,(H,12,17)(H,13,14)(H,15,16)(H,18,19)/t5-,6-/m0/s1

InChI key

RWSXRVCMGQZWBV-WDSKDSINSA-N

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

γ-Glu-Cys-Gly

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

May be used at 5-10 mM to elute glutathione S-transferase (GST) from glutathione agarose.

Biochem/physiol Actions

Endogenous antioxidant that plays a major role in reducing reactive oxygen species formed during cellular metabolism and the respiratory burst. Glutathione-S-transferase catalyzes the formation of glutathione thioethers with xenobiotics, leukotrienes, and other molecules that have an electrophilic center. Glutathione also forms disulfide bonds with cysteine residues in proteins. Via these mechanisms, it can have the paradoxical effect of reducing the efficacy of anti-cancer agents.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dalia Gritenaite et al.
Genes & development, 28(14), 1604-1619 (2014-07-18)
A key function of the cellular DNA damage response is to facilitate the bypass of replication fork-stalling DNA lesions. Template switch reactions allow such a bypass and involve the formation of DNA joint molecules (JMs) between sister chromatids. These JMs
Karen Shahbabian et al.
Nucleic acids research, 42(13), 8692-8704 (2014-07-12)
Messenger RNA (mRNA) localization is coupled to the translational repression of transcripts during their transport. It is still unknown if this coupling depends on physical interactions between translational control and mRNA localization machineries, and how these interactions are established at
Julien Viaud et al.
Nature communications, 5, 4080-4080 (2014-06-07)
PtdIns5P is a lipid messenger acting as a stress-response mediator in the nucleus, and known to maintain cell activation through traffic alterations upon bacterial infection. Here, we show that PtdIns5P regulates actin dynamics and invasion via recruitment and activation of
Y S L Lee et al.
Human reproduction (Oxford, England), 30(3), 543-552 (2015-01-09)
What is the relationship between cleavage stage embryo kinetics, blastocyst metabolism and subsequent embryo viability? Embryos cleaving faster at the first cleavage division resulted in blastocysts with a larger inner cell mass (ICM), higher glucose consumption, lower glycolytic rate, higher
Samir P Patel et al.
Experimental neurology, 257, 95-105 (2014-05-09)
Mitochondrial dysfunction is becoming a pivotal target for neuroprotective strategies following contusion spinal cord injury (SCI) and the pharmacological compounds that maintain mitochondrial function confer neuroprotection and improve long-term hindlimb function after injury. In the current study we evaluated the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service