Skip to Content
Merck
All Photos(1)

Key Documents

X70570

Sigma-Aldrich

Triton X-705 solution

70% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:

description

non-ionic

concentration

70% in H2O

technique(s)

ELISA: suitable
UV/Vis spectroscopy: suitable
protein purification: suitable
protein quantification: suitable

application(s)

detection

SMILES string

CC(C)(C)CC(C)(C)c1ccc(OCCOCCOCCOCCOCCOCCOCCO)cc1

InChI

1S/C28H50O8/c1-27(2,3)24-28(4,5)25-6-8-26(9-7-25)36-23-22-35-21-20-34-19-18-33-17-16-32-15-14-31-13-12-30-11-10-29/h6-9,29H,10-24H2,1-5H3

InChI key

HNLXNOZHXNSSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Russell M Morphew et al.
Veterinary parasitology, 191(3-4), 379-385 (2012-10-02)
Fasciola hepatica NP-40 released protein extract (FhNPE) exhibits potent Th1 immunosuppressive properties in vitro and in vivo. However, the protein composition of this active fraction, responsible for Th1 immune modulatory activity, has yet to be resolved. Therefore, FhNPE, a Nonidet
Halil İbrahim Ulusoy et al.
Talanta, 88, 516-523 (2012-01-24)
A new micelle-mediated separation and preconcentration method was developed for ultra-trace quantities of mercury ions prior to spectrophotometric determination. The method is based on cloud point extraction (CPE) of Hg(II) ions with polyethylene glycol tert-octylphenyl ether (Triton X-114) in the
Demirhan Citak et al.
Journal of AOAC International, 95(4), 1170-1175 (2012-09-14)
A cloud point extraction procedure was optimized for the separation and preconcentration of lead(II), cadmium(II), copper(II), and iron(III) ions in various water and canned food samples. The metal ions formed complexes with 2,6-diamino-4-phenyl-1,3,5-triazine that were extracted by surfactant-rich phases in
Ebru Ates et al.
Journal of AOAC International, 94(6), 1853-1862 (2012-02-11)
A cloud point extraction method is reported using LC/MS for the determination of regulated water-soluble food colors (Allura Red, Sunset Yellow, erythrosine, and tartrazine) and banned fat-soluble synthetic azo dyes (Sudan I, II, III, and IV; Red B; 7B; Black
Chunyapuk Kukusamude et al.
Journal of chromatography. A, 1351, 110-114 (2014-06-09)
Sample clean-up by cloud point phase separation and analysis by capillary electrophoresis with stacking was developed for quaternary ammonium herbicides (i.e., paraquat and diquat) in milk. For sample clean-up, a mixture of 845μL of milk sample, 5μL of 100mM phosphoric

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service