Skip to Content
Merck
All Photos(1)

Key Documents

W450001

Sigma-Aldrich

Choline chloride

FCC

Synonym(s):

(2-Hydroxyethyl)trimethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Cl)CH2CH2OH
CAS Number:
Molecular Weight:
139.62
FEMA Number:
4500
Beilstein:
3563126
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 117

form

powder, crystals or chunks

color

white

mp

302-305 °C (dec.) (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fishy

SMILES string

[Cl-].C[N+](C)(C)CCO

InChI

1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1

InChI key

SGMZJAMFUVOLNK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Choline chloride (ChCl) is a quaternary ammonium salt. The unusual solvent properties of eutectic mixtures of urea and choline chloride make it useful as a biodegradable ambient-temperature solvent for a number of applications.

Application


  • Profiling and optimized extraction of bioactive polyphenolic compounds from young, red-fleshed apple using eco-friendly deep eutectic solvents.: This research demonstrates the use of choline chloride-based deep eutectic solvents (DES) as a green alternative for extracting polyphenolic compounds from apples. The study highlights DESs effectiveness in yielding high-quality extracts, which could revolutionize natural product extraction processes in the food industry (Yu et al., 2024).

  • 17beta-estradiol induces hyperresponsiveness in guinea pig airway smooth muscle by inhibiting the plasma membrane Ca(2+)-ATPase.: This research utilizes choline chloride in physiological experiments to investigate the mechanisms through which estradiol affects calcium signaling in airway smooth muscle. The findings could lead to better understanding and treatment of asthma and other respiratory disorders (Romero-Martinez et al., 2024).

  • DES-ultrasonication treatment of cellulose nanocrystals and the reinforcement in carrageenan biocomposite.: In this study, choline chloride is used to create a novel biocomposite material, demonstrating its potential in enhancing the mechanical properties of biodegradable plastics. This could lead to new advancements in sustainable material science (Ramli et al., 2024).

  • A sensitive determination of Brilliant Blue FCF in some food samples using hydrophillic deep eutectic solvent-assisted magnetic nano gel-based dispersive solid phase microextraction prior to spectrophotometric analysis.: This innovative approach uses choline chloride in a deep eutectic solvent formulation to improve the detection of food dyes. The method promises greater efficiency and sensitivity, crucial for food safety and compliance (Erek et al., 2024).

Substrates

Choline acetyltransferase substrate

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of choline in milk, milk powder, and soy lecithin hydrolysates by flow injection analysis and amperometric detection with a choline oxidase based biosensor
Pati S, et al.
Journal of Agricultural and Food Chemistry, 52(15), 4638-4642 (2004)
Eutectic mixture of choline chloride/urea as a green solvent in synthesis of a coordination polymer:[Zn (O 3 PCH 2 CO 2)]? NH 4.
Liao JH, et al.
Inorganic Chemistry Communications, 8(4), 390-392 (2005)
Andrew P Abbott et al.
Chemical communications (Cambridge, England), (1)(1), 70-71 (2003-03-04)
Eutectic mixtures of urea and a range of quaternary ammonium salts are liquid at ambient temperatures and have interesting solvent properties.
Andrew P Abbott et al.
Physical chemistry chemical physics : PCCP, 11(21), 4269-4277 (2009-05-22)
Here we describe for the first time the electrolytic deposition of copper and copper composites from a solution of the metal chloride salt in either urea-choline chloride, or ethylene glycol-choline chloride based eutectics. We show that the deposition kinetics and
Green and moisture-stable Lewis acidic ionic liquids (choline chloride.xZnCl2) catalyzed protection of carbonyls at room temperature under solvent-free conditions.
Duan Z, et al.
Catalysis Communications, 7, 651-656 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service