Skip to Content
Merck
All Photos(1)

Documents

670189

Sigma-Aldrich

Cholin acetate

≥95.0% (T)

Synonym(s):

2-Hydroxyethyl-trimethylammonium acetate, COL Acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NO3
CAS Number:
Molecular Weight:
163.21
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (T)

form

solid

impurities

≤1.0% water

SMILES string

CC([O-])=O.C[N+](C)(C)CCO

InChI

1S/C5H14NO.C2H4O2/c1-6(2,3)4-5-7;1-2(3)4/h7H,4-5H2,1-3H3;1H3,(H,3,4)/q+1;/p-1

InChI key

QLKGUVGAXDXFFW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cholin acetate is a choline-derivative-based ionic liquid (IL), which can efficiently dissolve microcrystalline cellulose. This bioionic liquid can be prepared by reacting choline hydroxide with acetic acid.

Application

Cholin acetate in combination with glycerol, a hydrogen-bond donor forms a eutectic ionic liquid mixture, which is less viscous, biodegradable and also shows high lipase compatibility. This eutectic ionic liquid mixture can be a desirable solvent for the enzymatic production of biodiesel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

284.0 °F - closed cup

Flash Point(C)

140.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Low melting mixtures in organic synthesis-an alternative to ionic liquids?.
Ru? C and Konig B.
Green Chemistry, 14(11), 2969-2982 (2012)
Bio ionic liquids: room temperature ionic liquids composed wholly of biomaterials.
Fukaya Y, et al.
Green Chemistry, 9(11), 1155-1157 (2007)
Hua Zhao et al.
Organic & biomolecular chemistry, 9(6), 1908-1916 (2011-02-02)
The enzymatic preparation of biodiesel has been hampered by the lack of suitable solvents with desirable properties such as high lipase compatibility, low cost, low viscosity, high biodegradability, and ease of product separation. Recent interest in using ionic liquids (ILs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service