Skip to Content
Merck
All Photos(1)

Key Documents

S0360000

Selegiline hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

R-(−)-Deprenyl hydrochloride, (R)-(−)-N,α-Dimethyl-N-(2-propynyl)phenethylamine hydrochloride, R(−)-N-α-Dimethyl-N-2-propynyl-benzeneethanamine hydrochloride, Selegiline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N · HCl
CAS Number:
Molecular Weight:
223.74
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

selegiline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl[H].C[C@H](Cc1ccccc1)N(C)CC#C

InChI

1S/C13H17N.ClH/c1-4-10-14(3)12(2)11-13-8-6-5-7-9-13;/h1,5-9,12H,10-11H2,2-3H3;1H/t12-;/m1./s1

InChI key

IYETZZCWLLUHIJ-UTONKHPSSA-N

Gene Information

human ... MAOB(4129)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Selegiline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Selective MAO-B inhibitor; anti-Parkinsonian agent.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keiko Inaba-Hasegawa et al.
Journal of neural transmission (Vienna, Austria : 1996), 120(3), 435-444 (2012-09-13)
Type B monoamine oxidase (MAO-B) is proposed to be involved in the pathogenesis of neurodegenerative disorders, such as Parkinson's disease, through oxidative stress and synthesis of neurotoxins. MAO-B inhibitors, rasagiline and selegiline [(-)deprenyl], protect neuronal cells by direct intervention in
Retention rate of selegiline in early Parkinson's disease: a retrospective survey.
T Keränen et al.
International journal of clinical practice, 66(10), 1014-1014 (2012-09-22)
Wakako Maruyama et al.
Journal of neural transmission (Vienna, Austria : 1996), 120(1), 83-89 (2012-08-16)
Neuroprotection has been proposed in neurodegenerative disorders, such as Parkinson's and Alzheimer's diseases, to delay or halt disease progression or reverse neuronal deterioration. The inhibitors of type B monoamine oxidase (MAO), rasagiline and (-)deprenyl, prevent neuronal loss in cellular and
Rameshwar Singh et al.
Indian journal of experimental biology, 50(2), 101-109 (2012-06-08)
L-deprenyl is considered to protect against age-related cognitive deficits by improving long-term learning/memory in the aged brain. The CA1 and CA3 hippocampal areas are the sites at which initial learning and memory processes occur. Chronic deprenyl treatment significantly augmented the
Jorge A Zavala et al.
Journal of clinical neuroscience : official journal of the Neurosurgical Society of Australasia, 19(9), 1298-1299 (2012-06-23)
Pramipexole is a non-ergot dopamine agonist that is used frequently as a single therapy or in combination for the management of Parkinson's disease. Common side effects are daytime drowsiness, hypotension, hallucinations and compulsive behaviour. We describe a patient who developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service