Skip to Content
Merck
All Photos(1)

Key Documents

R1500000

Roxithromycin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Erythromycin 9-(-O-[2-methoxyethoxy]methyloxime)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H76N2O15
CAS Number:
Molecular Weight:
837.05
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

roxithromycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCC1OC(=O)C(C)C(OC2CC(C)(OC)C(O)C(C)O2)C(C)C(OC3OC(C)CC(C3O)N(C)C)C(C)(O)CC(C)C(=N/OCOCCOC)\C(C)C(O)C1(C)O

InChI

1S/C41H76N2O15/c1-15-29-41(10,49)34(45)24(4)31(42-53-21-52-17-16-50-13)22(2)19-39(8,48)36(58-38-32(44)28(43(11)12)18-23(3)54-38)25(5)33(26(6)37(47)56-29)57-30-20-40(9,51-14)35(46)27(7)55-30/h22-30,32-36,38,44-46,48-49H,15-21H2,1-14H3/b42-31+

InChI key

RXZBMPWDPOLZGW-FEMONOMJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Roxithromycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Semisynthetic erythromycin derivative. Impairs NADPH oxidase activation and alters translocation of its cytosolic components to the neutrophil membrane.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Guy et al.
Internal medicine journal, 41(10), 715-721 (2012-03-23)
Legionella species are a common cause of community-acquired pneumonia, infrequently complicated by cavitary disease. We describe Legionella pneumophila pneumonia and abscess formation in an immunosuppressed patient receiving corticosteroid therapy for metastatic breast carcinoma. The predisposing role of corticosteroids is discussed
Patorn Piromchai et al.
The Cochrane database of systematic reviews, (5)(5), CD008233-CD008233 (2011-05-13)
Chronic rhinosinusitis (CRS) is a common health problem which significantly affects quality of life. A wide range of medical and surgical therapies have been used to treat CRS. Both systemic and topical antibiotics are used with the aim of eliminating
Z Samra et al.
Journal of chemotherapy (Florence, Italy), 23(2), 77-79 (2011-05-17)
The in vitro activity of tetracycline, doxycycline, erythromycin, roxithromycin, clarithromycin, azithromycin, levofloxacin and moxifloxacin was tested against 63 clinical isolates of Ureaplasma urealyticum. The minimal inhibitory concentrations (MICs) and the minimal bactericidal concentrations (MBCs) were determined by the broth microdilution
Guy Rughani et al.
The Cochrane database of systematic reviews, 9(9), CD009536-CD009536 (2012-09-14)
Screening for abdominal aortic aneurysm (AAA) in selected groups is now performed in England, the USA and Sweden. Patients with aneurysms over 55 mm in diameter are generally considered for elective surgical repair. Patients with aneurysm diameters below or equal
A Markham et al.
Drugs, 48(2), 297-326 (1994-08-01)
Roxithromycin is a derivative of the macrolide antibacterial erythromycin with in vitro antibacterial activity resembling that of the parent compound. The drug has activity against some Staphylococcus spp., many Streptococcus spp., Moraxella (Branhamella) catarrhalis, Mycoplasma pneumoniae, Legionella pneumophila and Chlamydia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service