Skip to Content
Merck
All Photos(2)

Documents

97599

Supelco

Salvianolic acid A

analytical standard

Synonym(s):

R)-α-[[(2E)-3-[2-[(1E)-2-(3,4-Dihydroxyphenyl)ethenyl]-3,4-dihydroxyphenyl]-1-oxo-2-propen-1-yl]oxy]-3,4-dihydroxybenzenepropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22O10
CAS Number:
Molecular Weight:
494.45
Beilstein:
2660108
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC1=CC=C(C(/C=C/C2=CC(O)=C(C=C2)O)=C1O)/C=C/C(O[C@H](CC3=CC(O)=C(C=C3)O)C(O)=O)=O

InChI

1S/C26H22O10/c27-18-7-2-14(11-21(18)30)1-6-17-16(4-9-20(29)25(17)33)5-10-24(32)36-23(26(34)35)13-15-3-8-19(28)22(31)12-15/h1-12,23,27-31,33H,13H2,(H,34,35)/b6-1+,10-5+/t23-/m1/s1

InChI key

YMGFTDKNIWPMGF-UCPJVGPRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Salvianolic acid A is a phenolic acid, isolated from Danshen, a traditional Chinese medicine, commonly used for the treatment of bone loss, chronic renal failure, hepatocirrhosis, hepatitis, coronary heart disease and cerebrovascular disease.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Salvianolic acid A may be used as an analytical reference standard for the quantification of the analyte in Chinese medicinal preparations using liquid chromatography

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: salvia

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kai Wang et al.
Nutrients, 10(6) (2018-06-21)
Salvianolic acid A (SAA) is an active phenolic acid derived from Salvia miltiorrhiza Bunge (Danshen). To explore whether SAA has a therapeutic effect against inflammatory bowel disease (IBD), an acute colitis model was induced in rats by administering 3% dextran
Ting Yu et al.
Computational biology and chemistry, 74, 273-285 (2018-04-22)
The rhizome of Salvia miltiorrhiza has emerged as a rich source of natural therapeutic agents, and its several compounds are supposed to exhibit favorable effects on Alzheimer's disease (AD). The present work investigate the anti-AD potentials of 12 tanshinones, three
Meriem Taghouti et al.
Antioxidants (Basel, Switzerland), 9(1) (2020-01-08)
Thymusmastichina, also called mastic thyme or Spanish marjoram, is endemic to the Iberian Peninsula, where it is widely used in folk medicine especially for treating digestive and respiratory systems disorders, and as a condiment to season olives. This work describes
Chunyang Xu et al.
Oxidative medicine and cellular longevity, 2020, 6431459-6431459 (2020-03-19)
Oxidative stress has been recognized as the contributor to diabetic peripheral neuropathy (DPN). Antioxidant strategies have been most widely explored; nevertheless, whether antioxidants alone prevent DPN still remains inconclusive. In the present study, we established an in vitro DPN cell
Yipeng Ma et al.
International journal of antimicrobial agents, 53(2), 128-136 (2018-08-01)
Human enterovirus 71 (EV-A71) infections cause a wide array of diseases ranging from diarrhoea and rashes to hand-foot-and-mouth disease and, in rare cases, severe neurological disorders. No specific antiviral drug therapy is currently available. Extracts from 75 Chinese medicinal plants

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service