Skip to Content
Merck
All Photos(1)

Key Documents

PHR1713

Supelco

Proguanil hydrochloride

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Proguanil hydrochloride, Chlorguanide, N1-(4-Chlorophenyl)-N5-isopropylbiguanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16ClN5·HCl
CAS Number:
Molecular Weight:
290.19
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 297
traceable to USP 1568040

API family

proguanil

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

Cl.CC(C)NC(=N)NC(=N)Nc1ccc(Cl)cc1

InChI

1S/C11H16ClN5.ClH/c1-7(2)15-10(13)17-11(14)16-9-5-3-8(12)4-6-9;/h3-7H,1-2H3,(H5,13,14,15,16,17);1H

InChI key

SARMGXPVOFNNNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Proguanil hydrochloride is a biguanide compound, widely used as an antimalarial drug. It exhibits sporontocidal activity, thereby rendering the gametocytes non-infective to the mosquito vector.
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Application

Proguanil hydrochloride may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using high-performance liquid chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Chlorguanide (proguanil) is combined with atovaquone for malaria prophylaxis. The two compounds act synergistically to inhibit the plasmodial dihydrofolate reductase (DHFR) and interrupt the electron transport chain. Mutations in DHFR account for the development of resistant strains.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA7301 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guidelines for the Treatment of Malaria (2010)
Dual-mode gradient HPLC procedure for the simultaneous determination of chloroquine and proguanil
Paci.A, et al.
Journal of Pharmaceutical and Biomedical Analysis, 27(1-2), 1-7 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service