Skip to Content
Merck
All Photos(1)

Key Documents

G7048

Sigma-Aldrich

Proguanil hydrochloride

≥95% (HPLC)

Synonym(s):

Chlorguanide, N1-(4-Chlorophenyl)-N5-isopropylbiguanide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16ClN5·HCl
CAS Number:
Molecular Weight:
290.19
EC Number:
MDL number:
UNSPSC Code:
51101906
PubChem Substance ID:
NACRES:
NA.77

Assay

≥95% (HPLC)

form

solid

storage condition

desiccated

solubility

acetonitrile: water: ~1 mg/mL (60/40)

originator

AstraZeneca

storage temp.

2-8°C

SMILES string

Cl.CC(C)NC(=N)NC(=N)Nc1ccc(Cl)cc1

InChI

1S/C11H16ClN5.ClH/c1-7(2)15-10(13)17-11(14)16-9-5-3-8(12)4-6-9;/h3-7H,1-2H3,(H5,13,14,15,16,17);1H

InChI key

SARMGXPVOFNNNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Proguanil (chlorguanide) may be used in anti-parasitic protozoan drug development to study its pharmacokinetics, metabolism, safety, efficacy and methods of delivery as an antimalarial drug.

Biochem/physiol Actions

Chlorguanide (proguanil) is combined with atovaquone for malaria prophylaxis. The two compounds act synergistically to inhibit the plasmodial dihydrofolate reductase (DHFR) and interrupt the electron transport chain. Mutations in DHFR account for the development of resistant strains.

Features and Benefits

This compound was developed by AstraZeneca. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carine Van Malderen et al.
Malaria journal, 11, 139-139 (2012-05-02)
Malaria is a leading cause of mortality, particularly in sub-Saharan African children. Prompt and efficacious treatment is important as patients may progress within a few hours to severe and possibly fatal disease. Chlorproguanil-dapsone-artesunate (CDA) was a promising artemisinin-based combination therapy
Miriam K Laufer et al.
PloS one, 7(8), e42284-e42284 (2012-08-23)
The predominance of chloroquine-susceptible falciparum malaria in Malawi more than a decade after chloroquine's withdrawal permits contemplation of re-introducing chloroquine for targeted uses. We aimed to compare the ability of different partner drugs to preserve chloroquine efficacy and prevent the
Allan Pamba et al.
Blood, 120(20), 4123-4133 (2012-09-21)
Drug-induced acute hemolytic anemia led to the discovery of G6PD deficiency. However, most clinical data are from isolated case reports. In 2 clinical trials of antimalarial preparations containing dapsone (4,4'-diaminodiphenylsulfone; 2.5 mg/kg once daily for 3 days), 95 G6PD-deficient hemizygous
S Looareesuwan et al.
The American journal of tropical medicine and hygiene, 60(4), 533-541 (1999-05-29)
The continuing spread of drug-resistant malaria emphasizes the need for new antimalarial drugs. Atovaquone is a broad-spectrum antiprotozoal drug with a novel mechanism of action, via inhibition of parasite mitochondrial electron transport, and a favorable safety profile. Early studies with
Sarah Donegan et al.
Statistics in medicine, 31(29), 3840-3857 (2012-07-13)
Mixed treatment comparison (MTC) meta-analysis allows several treatments to be compared in a single analysis while utilising direct and indirect evidence. Treatment by covariate interactions can be included in MTC models to explore how the covariate modifies the treatment effects.

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service