Skip to Content
Merck
All Photos(2)

Key Documents

P1254

Sigma-Aldrich

Brij® L23

suitable for Stein-Moore chromatography

Synonym(s):

Brij 35, C12E23, Polyoxyethylene (23) lauryl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

non-ionic

Quality Level

form

solid

mol wt

micellar avg mol wt 48,000
estimated mol wt 1198

aggregation number

20-40

CMC

91 μM

transition temp

cloud point >100 °C

HLB

16.9

suitability

suitable for Stein-Moore chromatography

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C58H118O24/c1-2-3-4-5-6-7-8-9-10-11-13-60-15-17-62-19-21-64-23-25-66-27-29-68-31-33-70-35-37-72-39-41-74-43-45-76-47-49-78-51-53-80-55-57-82-58-56-81-54-52-79-50-48-77-46-44-75-42-40-73-38-36-71-34-32-69-30-28-67-26-24-65-22-20-63-18-16-61-14-12-59/h59H,2-58H2,1H3

InChI key

IEQAICDLOKRSRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Non-ionic detergent. Useful for the extraction of membrane proteins. Typically used in the range of 0.1-1%.

Legal Information

Brij is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

>300.2 °F - open cup

Flash Point(C)

> 149 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Yamamoto et al.
Biochimica et biophysica acta, 790(3), 208-218 (1984-11-09)
Two forms of an acid proteinase have been purified from human erythrocyte membranes by a simple method involving selective extraction with 0.5% Brij 35, affinity chromatography on pepstatin A-Sepharose 4B, and chromatography on Sephacryl S-200 and DEAE-Sephadex G-100. One species
T Tobimatsu et al.
Archives of biochemistry and biophysics, 347(1), 132-140 (1997-11-05)
Recombinant adenosylcobalamin-dependent diol dehydratase of Klebsiella oxytoca overexpressed in Escherichia coli was purified to homogeneity. The enzyme has a low solubility and was extracted from the crude membrane fraction with 1% Brij 35 in a high recovery. Subsequent chromatography on
Jacomina W van den Esschert et al.
Annals of surgery, 255(2), 311-318 (2012-01-14)
To compare the safety and hypertrophy response after portal vein embolization (PVE) using 2 absorbable and 3 permanent embolization materials. Portal vein embolization is used to increase future remnant liver volume preoperatively. Application of temporary, absorbable embolization materials could be
Jun Koizumi et al.
AJR. American journal of roentgenology, 199(1), 200-207 (2012-06-27)
Balloon-occluded retrograde transvenous obliteration has been traditionally based on liquid sclerotherapy. However, overdose and systemic spillage of liquid sclerosant can cause severe complications, such as hemolysis, which lead to hemoglobinuria, allergy, acute respiratory distress syndrome, and other disorders. The purpose
Otte Brosjö et al.
Acta orthopaedica, 84(5), 502-505 (2013-11-01)
Recent data suggest that percutaneous sclerotherapy is a safe alternative to surgery for treatment of aneurysmal bone cysts (ABCs). We present our experience of this method. We retrospectively analyzed data from 38 consecutive patients treated with repeated injections of polidocanol.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service