Skip to Content
Merck
All Photos(1)

Key Documents

P0532

Sigma-Aldrich

Phthaldialdehyde Reagent

Solution Complete

Synonym(s):

OPA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21

Quality Level

storage temp.

2-8°C

SMILES string

[H]C(=O)c1ccccc1C([H])=O

InChI

1S/C8H6O2/c9-5-7-3-1-2-4-8(7)6-10/h1-6H

InChI key

ZWLUXSQADUDCSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precolumn derivatization reagent for primary amines and amino acids. The fluorescent derivative can be detected by reverse-phase HPLC. The reaction requires OPA, primary amine and a sulfhydryl. In the presence of excess sulfhydryl, amines can be quantitated. In the presence of excess amine, sulfhydryls can be quantitated.

Other Notes

Contains 1 mg o-phthaldialdehyde (P0657) per mL solution with 2-mercaptoethanol as the sulfhydryl moiety.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B - Skin Sens. 1

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silke B Lohan et al.
Antioxidants (Basel, Switzerland), 9(6) (2020-06-25)
The daily consumption of tobacco products leads to a boost in free radical production in tissues, promoting the risk for malignancies, metabolic alterations and chronic-inflammatory diseases. This study aimed to broaden the knowledge of the status of the antioxidative (AO)
Gianni Pala et al.
Expert review of clinical immunology, 9(3), 227-234 (2013-03-01)
This study summarizes and reviews the available health information on ortho-phthalaldehyde (OPA), a recently introduced and widespread disinfectant for heat-sensitive medical equipment, particularly focusing on its possible immunological effects in the healthcare setting. OPA properties derived from laboratory and clinical
Jong-Suk Song et al.
Investigative ophthalmology & visual science, 53(10), 6164-6170 (2012-08-18)
We evaluated whether viscoelastics have protective effects on the corneal endothelial cell damage in a toxic anterior segment syndrome (TASS) animal model depending on the types of viscoelastics. A TASS animal model was established with an injection of 0.1 mL
A Pappa-Louisi et al.
Talanta, 85(4), 2241-2245 (2011-08-30)
In an effort to enhance complex mixture separations by using small amounts of a homologous series of alkanols as additives in the mobile phases, it was proposed an optimization algorithm based on a sixth-parameter retention model. This model considers simultaneously
M M Ayyash et al.
Journal of food science, 77(8), M490-M498 (2012-07-20)
The effect of NaCl substitution with KCl at different pH levels (6.0, 5.5, and 5.0) and salt concentrations on proteinase activities of cell-free and supernatant of Lactobacillus delbrueckii ssp. bulgaricus 11824 (L. bulgaricus) and Streptococcus thermophilus MS (ST) was investigated.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service