Skip to Content
Merck
All Photos(1)

Key Documents

O0270000

Oxybuprocaine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Benoxinate hydrochloride, 2-(Diethylamino)ethyl 4-amino-3-butoxybenzoate hydrochloride, 4-Amino-3-butoxybenzoic acid diethylaminoethyl ester, Oxybuprocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H28N2O3 · HCl
CAS Number:
Molecular Weight:
344.88
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

oxybuprocaine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].CCCCOc1cc(ccc1N)C(=O)OCCN(CC)CC

InChI

1S/C17H28N2O3.ClH/c1-4-7-11-21-16-13-14(8-9-15(16)18)17(20)22-12-10-19(5-2)6-3;/h8-9,13H,4-7,10-12,18H2,1-3H3;1H

InChI key

PRGUDWLMFLCODA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Oxybuprocaine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cirous Dehghani et al.
Investigative ophthalmology & visual science, 55(5), 3195-3199 (2014-04-26)
We examined the age-dependent alterations and longitudinal course of subbasal nerve plexus (SNP) morphology in healthy individuals. Laser-scanning corneal confocal microscopy, ocular screening, and health and metabolic assessment were performed on 64 healthy participants at baseline and at 12-month intervals
Franziska Bucher et al.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie, 252(7), 1119-1126 (2014-05-31)
To analyze potential alterations in corneal nerve morphology and function in different stages of Fuchs' endothelial corneal dystrophy (FECD). Thirty eyes with FECD underwent in vivo confocal microscopy using the Heidelberg Retina Tomograph II (HRT II; Heidelberg Engineering, Heidelberg, Germany)
Kumiko Kato
Veterinary ophthalmology, 17(6), 417-425 (2014-02-13)
To compare the intraocular pressure (IOP) measurements from two applanation tonometers (Tono-Pen XL(®) and AccuPen(®) in normal and diseased canine eyes (including corneal disease, cataract, glaucoma, uveitis, and noninflammatory intraocular disease) and evaluate the associations among the central corneal thickness
Leilei Zou et al.
Molecular vision, 20, 977-987 (2014-07-16)
Scleral remodeling is an important mechanism underlying the development of myopia. Atropine, an antagonist of G protein-coupled muscarinic receptors, is currently used as an off-label treatment for myopia. Regulator of G-protein signaling 2 (RGS2) functions as an intracellular selective inhibitor
Nakhleh E Abu-Yaghi et al.
International journal of ophthalmology, 7(6), 1017-1021 (2014-12-30)
To document the indications, safety and possible complications of bilateral same-session intravitreal anti-vascular endothelial growth factor (VEGF) injections performed in the ophthalmic operating room. A retrospective case series study. Consecutive records of seventy four patients receiving simultaneous bilateral intravitreal injections

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service