Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Nicotinic acid
Synonym(s):
3-Picolinic acid, Niacin, Pellagra preventive factor, Pyridine-3-carboxylic acid, Vitamin B
3
, Nicotinic acid, Nicotinic Acid
Linear Formula:
3-(COOH)-C
5
H
4
N
CAS Number:
59-67-6
Molecular Weight:
123.11
EC Number:
200-441-0
Products
Product Category
vitamins (5)
Brand
Millipore (1)
Sigma-Aldrich (5)
Biological Source
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Technique
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
72309
Nicotinic acid,
≥99.5% (HPLC)
Expand
View Pricing
N0761
Nicotinic acid,
BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Expand
View Pricing
N4126
Nicotinic acid,
≥98%
Expand
View Pricing
8.18714
Nicotinic acid,
for synthesis
Expand
View Pricing
481918
Nicotinic acid,
A component of coenzymes NAD+ and NADP+.
Expand
View Pricing
N5410
Nicotinic acid,
meets USP testing specifications
Expand
View Pricing
21-1430
Nicotinic acid,
SAJ special grade, ≥99.5%
Expand
View Pricing