Skip to Content
Merck
All Photos(3)

Key Documents

M8179

Sigma-Aldrich

Manganese sulfate monohydrate

meets USP testing specifications

Synonym(s):

Manganese(II) sulfate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnSO4 · H2O
CAS Number:
Molecular Weight:
169.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Quality Level

Agency

USP/NF
meets USP testing specifications

form

powder

application(s)

pharmaceutical (small molecule)

SMILES string

O.[Mn++].[O-]S([O-])(=O)=O

InChI

1S/Mn.H2O4S.H2O/c;1-5(2,3)4;/h;(H2,1,2,3,4);1H2/q+2;;/p-2

InChI key

ISPYRSDWRDQNSW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Promoting nitrogen conversion in aerobic biotransformation of swine slurry with the co-application of manganese sulfate and biochar</strong>: This study explores the synergistic effects of manganese sulfate monohydrate and biochar on nitrogen conversion processes in swine slurry management, highlighting its potential to enhance biotransformation efficiency in agricultural waste management (Wu et al., 2024).</li>
<li><strong>Multi-Omics Analysis of the Effects of Soil Amendment on Rapeseed (Brassica napus L.) Photosynthesis under Drip Irrigation with Brackish Water</strong>: This study indicates the broader agricultural applications of manganese sulfate monohydrate in enhancing crop photosynthesis and overall yield, particularly under challenging irrigation conditions (Li et al., 2024).</li>
<li><strong>The influence of steroidal implants and manganese sulfate supplementation on growth performance, trace mineral status, hepatic gene expression, hepatic enzyme activity, and circulating metabolites in feedlot steers</strong>: This research demonstrates the beneficial effects of manganese sulfate monohydrate on animal health, specifically in boosting growth performance and mineral status in livestock (Smerchek et al., 2024).</li>
</ul>

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - STOT RE 2 Inhalation

Target Organs

Brain

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rahim Khabaz
Radiation protection dosimetry, 152(4), 400-405 (2012-05-26)
In this study, an approximate symmetric cylindrical manganese bath system with equal diameter and height was appraised using a Monte Carlo simulation. For nine sizes of the tank filled with MnSO(4).H(2)O solution of three different concentrations, the necessary correction factors
Ramon Frick et al.
Toxicology letters, 205(2), 163-172 (2011-06-15)
Due to their physicochemical characteristics, metal oxide nanoparticles (NPs) interact differently with cells compared to larger particles or soluble metals. Oxidative stress and cellular metal uptake were quantified in rat type II alveolar epithelial cells in culture exposed to three
Zhehui Weng et al.
Inorganic chemistry, 48(18), 8703-8708 (2009-08-25)
A new glycine-templated manganese(II) sulfate, (C(2)NO(2)H(5))MnSO(4) (1), has been solvothermally synthesized and characterized crystallographically and magnetically. Crystal data for 1: monoclinic, space group P2(1)/m, a = 4.8884(10) A, b = 7.8676(14) A, c = 8.0252(15) A, beta = 106.524(2) degrees
A P Marreilha dos Santos et al.
Brain research, 1236, 200-205 (2008-08-30)
Manganese (Mn) is an essential trace element required for ubiquitous enzymatic reactions. Chronic overexposure to this metal may, however, promote potent neurotoxic effects. The mechanism of Mn toxicity is not well established, but several studies indicate that oxidative stress and
Long Peng et al.
Journal of colloid and interface science, 377(1), 13-17 (2012-04-21)
A facile single-source precursor method has been applied for the selective synthesis of MnS nanocrystals (NCs) with well-defined shapes and crystal structures such as hexapod, octahedral, hexagonal shaped α-MnS NCs, and pencil-shaped γ-MnS NCs. The effects of the composition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service