Skip to Content
Merck
All Photos(1)

Documents

P4780

Sigma-Aldrich

TWEEN® 80

suitable for cell culture, suitable for insect cell culture, viscous liquid

Synonym(s):

POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161700
NACRES:
NA.25

description

non-ionic

Quality Level

form

viscous liquid

mol wt

average mol wt ~1310

composition

Oleic acid, ≥58.0% (balance primarily linoleic, palmitic, and stearic acids)

technique(s)

cell culture | insect: suitable
cell culture | mammalian: suitable

CMC

0.012 mM

density

1.06 g/mL at 25 °C (lit.)
1.07 g/cm3 at 25 °C (lit.)

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TWEEN 80 is a polyethylene sorbitol ester and has a molecular weight of 1.31 kDa. It is useful as a stabilizer and emulsifier commercially. It is known to be used in ice-cream for its drying property.

Application

Non-ionic detergent used for selective protein extraction and isolation of nuclei from mammalian cell lines.
TWEEN® 80 has been used:
  • in Listeria monocytogenes enumeration using filtration method
  • as a hydrophilic emulsifier in the preparation of W1/O/W2 emulsion
  • as a component of 7H9 plus oleic albumin dextrose catalase (OADC) in Mycobacterium Bovis bacillus Calmette–Guerin (BCG) Pasteur strain culturing

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ice Cream, 86-86 (2003)
Low-fat Gouda cheese made from bovine milk-olive oil emulsion: physicochemical and sensory attributes
Felfoul I, et al.
Journal of Food Science and Technology, 52(10), 6749-6755 (2015)
A contribution to the improvement of Listeria monocytogenes enumeration in cold-smoked salmon
Besse NG, e al.
International Journal of Food Microbiology, 91(2), 119-127 (2004)
Fats in Food Technology (2014)
Differential requirements for L-citrulline and L-arginine during antimycobacterial macrophage activity
Rapovy SM, et al.
Journal of Immunology, 195(7), 3293-3300 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service