Skip to Content
Merck
All Photos(1)

Key Documents

G0050000

Galactose

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-(+)-Galactose, Galactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724619
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

galactose

manufacturer/tradename

EDQM

mp

168-170 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6-/m0/s1

InChI key

GZCGUPFRVQAUEE-KCDKBNATSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Galactose EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhuoran Wang et al.
PloS one, 9(7), e102118-e102118 (2014-07-11)
Pten is a tumor suppressor gene regulating many cellular processes, including growth, adhesion, and apoptosis. In the aim of investigating the role of Pten during mammary gland development and lactation of dairy cows, we analyzed Pten expression levels in the
Michelle P Christie et al.
PloS one, 9(4), e95024-e95024 (2014-04-17)
Glycosylation of biopharmaceuticals can mediate cell specific delivery by targeting carbohydrate receptors. Additionally, glycosylation can improve the physico-chemical (drug-like) properties of peptide based drug candidates. The main purpose of this study was to examine if glycosylation of the peptide enkephalin
Yanhua Yao et al.
PLoS genetics, 11(1), e1004968-e1004968 (2015-01-30)
Elevated proteasome activity extends lifespan in model organisms such as yeast, worms and flies. This pro-longevity effect might be mediated by improved protein homeostasis, as this protease is an integral module of the protein homeostasis network. Proteasomes also regulate cellular
Guan-Ming Qi et al.
Endocrinology, 155(6), 2254-2265 (2014-04-02)
Previous studies have indicated that adiponectin (APN) protects against cardiac remodeling, but the underlying mechanism remains unclear. The present study aimed to elucidate how APN regulates inflammatory responses and cardiac fibrosis in response to angiotensin II (Ang II). Male APN
Lu Han et al.
Biomaterials, 44, 111-121 (2015-01-27)
Multifunctional nanocomplexes (NCs) consisting of urocanic acid-modified galactosylated trimethyl chitosan (UA-GT) conjugates as polymeric vectors, poly(allylamine hydrochloride)-citraconic anhydride (PAH-Cit) as charge-reversible crosslinkers, and vascular endothelial growth factor (VEGF) siRNA as therapeutic genes, were rationally designed to simultaneously overcome the extracellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service