Skip to Content
Merck
All Photos(8)

Key Documents

G0750

Sigma-Aldrich

D-(+)-Galactose

D-(+)-Galactose

≥99% (HPLC)

Synonym(s):

Galactose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein:
1724619
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

biological source

bovine (Ruminant- Cow, Ox, Buffalo)

Quality Level

Assay

≥99% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

≤0.1% glucose

color

white

useful pH range

5.0-7 (25 °C, 180 g/L)

mp

168-170 °C (lit.)

solubility

H2O: soluble 100 mg/mL, clear, colorless

storage temp.

room temp

SMILES string

OC[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2/t3-,4+,5+,6-/m0/s1

InChI key

GZCGUPFRVQAUEE-KCDKBNATSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Galactose has been used:
  • as a component of galactosyltransferase labeling buffer.[1]
  • as a supplement in MRS broth for the growth of thermophilic lactobacilli[2]
  • to induce the expression of uncoupling protein (UCP) in yeast transformants[3]

Biochem/physiol Actions

Galactose is a simple monosaccharide that serves as an energy source and as an essential component of glycolipids and glycoproteins. Galactose contributes to energy metabolism via its conversion to glucose by the enzymes that constitute the Leloir pathway. Defects in the genes encoding these proteins lead to the metabolic disorder galactosemia.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D L Murdza-Inglis et al.
The Journal of biological chemistry, 269(10), 7435-7438 (1994-03-11)
The uncoupling protein is one of a family of mitochondrial transport proteins involved in energy metabolism. It dissipates oxidative energy to generate heat, either by catalyzing proton transport directly or by catalyzing fatty acid anion transport, thus enabling fatty acids
K W Turner et al.
Applied and environmental microbiology, 45(6), 1932-1934 (1983-06-01)
The ability to ferment galactose is a major characteristic which can be used to differentiate Lactobacillus helveticus (galactose positive) from Lactobacillus lactis and Lactobacillus bulgaricus (galactose negative). In milk cultures, galactose-positive strains produced d- and l-lactic acid with little galactose
Inge E Krabbendam et al.
Cell death & disease, 11(4), 263-263 (2020-04-25)
Metabolic flexibility is an essential characteristic of eukaryotic cells in order to adapt to physiological and environmental changes. Especially in mammalian cells, the metabolic switch from mitochondrial respiration to aerobic glycolysis provides flexibility to sustain cellular energy in pathophysiological conditions.
Zu-Hong He et al.
Redox biology, 28, 101364-101364 (2019-11-16)
Inflammation is a self-defense response to protect individuals from infection and tissue damage, but excessive or persistent inflammation can have adverse effects on cell survival. Many individuals become especially susceptible to chronic-inflammation-induced sensorineural hearing loss as they age, but the
Luke E Formosa et al.
Cell reports, 31(3), 107541-107541 (2020-04-23)
Mitochondrial complex I harbors 7 mitochondrial and 38 nuclear-encoded subunits. Its biogenesis requires the assembly and integration of distinct intermediate modules, mediated by numerous assembly factors. The mitochondrial complex I intermediate assembly (MCIA) complex, containing assembly factors NDUFAF1, ECSIT, ACAD9

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-only review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service